Skip to Content
Merck
All Photos(3)

Key Documents

O2881

Sigma-Aldrich

Oxybutynin chloride

≥98% (TLC), powder

Synonym(s):

α-Phenylcyclohexaneglycolic acid 4-(diethylamino)-2-butynyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO3 · HCl
CAS Number:
Molecular Weight:
393.95
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

originator

Johnson & Johnson

SMILES string

Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2

InChI

1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H

InChI key

SWIJYDAEGSIQPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Muscarinic acetylcholine receptor antagonist.
Muscarinic acetylcholine receptor antagonist; inhibits proliferation of bladder smooth muscle cells, perhaps by downregulation of growth promoting genes.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K-S Lee et al.
International journal of clinical practice, 66(7), 663-670 (2012-06-16)
Standardised traditional outcome measures may fail to address factors that are important to patients and address irrelevant factors. Aim of this study was to assess patient-reported goals and goal achievement (GA) in the antimuscarinic treatment for overactive bladder (OAB) patients.
Lorne E Aaron et al.
Current medical research and opinion, 28(8), 1369-1379 (2012-07-10)
Evaluate patient and physician satisfaction with a novel formulation of a once-daily controlled-release (CR) oxybutynin (Uromax*) 15-mg tablet as both the initial and maintenance dose in elderly and non-elderly patients with overactive bladder (OAB). Patients not on anticholinergic treatment for
Adding to the evidence base: application drying time for oxybutynin chloride topical gel.
Cynthia M Sublett
Urologic nursing, 32(5), 270-271 (2013-03-12)
V Schuler et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(10), 947-951 (2012-07-25)
A broad spectrum of synthetic agents is available for the treatment of overactive bladder. Anti-cholinergic drugs show a poor compliance due to side effects. There is an increasing use of plant extracts in medicine. We have therefore investigated the inhibitory
Jessica T Casey et al.
The Journal of urology, 188(4 Suppl), 1516-1520 (2012-08-23)
Abnormal bladder function following posterior urethral valve ablation can lead to deleterious effects on renal function and urinary continence. We performed a pilot study to determine if bladder dysfunction could be ameliorated by the early administration of oxybutynin. We enrolled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service