NIST3299
Ground Turmeric (Curcuma longa L.) Rhizome
NIST®SRM®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
form
powder
packaging
pkg of 15 g (5 pouches of 3g each)
manufacturer/tradename
NIST®
application(s)
agriculture
storage temp.
15-25°C
Related Categories
General description
A single unit of SRM 3299 comprises five pouches, each containing about 3 g of turmeric rhizome. For more information, please refer to the SDS and COA.
SRM 3299_cert
SRM 3299_SDS
SRM 3299_cert
SRM 3299_SDS
Application
This Standard Reference Material (SRM) is intended for validating analytical methods used to determine curcuminoids and elements in ground turmeric (Curcuma longa L.) rhizomes and similar matrices.
Features and Benefits
- NIST certified values are reported on a dry-mass basis, for curcuminoids and elements that are metrologically traceable to the SI unit of mass fraction and determined using inductively coupled plasma mass spectrometry (ICP-MS).
- Expiration details, instructions for use, storage, and handling are provided on the NIST certificate.
Other Notes
Certified for the analytes listed below.
Bisdemethoxycurcumin, Desmethoxycurcumin, Curcumin, Arsenic (As), Cadmium (Cd), Lead (Pb)
See certificate for values and more details at nist.gov/SRM.
Bisdemethoxycurcumin, Desmethoxycurcumin, Curcumin, Arsenic (As), Cadmium (Cd), Lead (Pb)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service