Skip to Content
Merck
All Photos(2)

Key Documents

C7727

Sigma-Aldrich

Curcumin

≥94% (curcuminoid content), ≥80% (Curcumin)

Synonym(s):

(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
Colour Index Number:
75300
Beilstein:
2306965
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

vapor density

13 (vs air)

Quality Level

Assay

≥80% (Curcumin)
≥94% (curcuminoid content)

form

powder

mp

175 °C

solubility

ethanol: 1 mg/mL
DMSO: >11 mg/mL
0.5 M NaOH: soluble (then immediately diluted in PBS)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Curcumin is a yellow colored polyphenol obtained from the rhizome of Curcuma longa L. It is also known as diferuloyl methane, a symmetric molecule with a seven carbon linker connecting two o-methoxy phenolic groups containing aromatic rings.[1] Curcumin is synthesized by Pabon′s reaction using vanillin and acetyl acetone in the presence of boric oxide.[2] It constitutes an important part of various diets and herbal medicines, especially in the Asian continent for its various salubrious effects on human health.[3][4]

Application

Curcumin has been used:
  • to study its in vitro effect on the inflammation and expression of tumour factor in patients with chronic kidney diseases[5]
  • to determine its influence when used in combination with high frequency ultrasound on human cervical carcinoma cells[6]
  • to study the possibility of using magnetic purification of curcumin from Curcuma longa by surface active maghemite nanoparticles (SAMNs)[7]
  • to examine its effect on the amyloid-β peptide levels along with the maturation of amyloid-β precursor proteins in various cell lines and primary cortical neurons in mouse[8]
  • as a supplement to evaluate its anti-carcinogenic and anti-apoptotic effect on cancer cells and cardiomyocytes[9]
  • to test its anti-inflammatory effect in human promyelocytic leukemia cell line, the HL-60[10]

Biochem/physiol Actions

A natural phenolic compound responsible for the yellow color of turmeric. Potent anti-tumor agent that acts against highly diverse tumor-specific pathways. Anti-inflammatory and anti-oxidant. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cyclooxygenase (COX-2) and 5-lipoxygenase (5-LOX).
A natural phenolic compound. Potent anti-tumor agent having anti-inflammatory and anti-oxidant properties. Curcumin has been cited as a potential chemopreventive agent, in addition to its chemotherapeutic activity. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cycloxygenase and lipoxygenase. Easily penetrates into the cytoplasm of cells, accumulating in membranous structures such as plasma membrane, endoplasmic reticulum and nuclear envelope.
Curcumin is postulated to have anti-depressant, anti-stress and neuroprotective effects on humans and other animals.[4] Additionally, it also shows anti-fungal, antiviral, anti-microbial, chemosensitizing, radiosensitizing, and wound healing activities.[11]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of turmeric extracts on inflammatory mediator production
Lantz RC, et al.
Phytomedicine, 12(6-7), 445-452 (2005)
Revisiting curcumin chemistry part I: A new strategy for the synthesis of curcuminoids
Rao EV and Sudheer P
Indian Journal of Pharmaceutical Sciences, 73(3), 262-262 (2011)
The chemistry of curcumin: from extraction to therapeutic agent
Priyadarsini K
Molecules (Basel), 19(12), 20091-20112 (2014)
Gizem Calibasi-Kocal et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 24(4), 1482-1487 (2019-10-28)
Curcumin is a natural phytopolyphenol compound isolated from the root of turmeric (Curcuma longa) and possesses a wide range of biological properties. The purpose of this study was to evaluate the antiproliferative, wound healing, anti-invasive and anti-migrative effects of curcumin
Assessment of herb-drug synergy to combat doxorubicin induced cardiotoxicity
Jain A and Rani V
Life Sciences, 205(3), 97-106 (2018)

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. how does this product relates to commercially available curcumin available as supplement?

    1 answer
    1. This product is the same compound as those sold commercially, however this item is intended for research use only. It is not suitable for human consumption.

      Helpful?

  4. Hi, may I know if I can store curcumin (c7727) in ethanol in 4 degree celcius. If yes, for how long? Thank you

    1 answer
    1. The storage temperature of Curcumin stock solutions has not been determined. Please see the links below referencing the solution storage conditions of Curcumin:
      Physical and Chemical Stability of Curcumin in Aqueous Solutions and Emulsions
      https://pubs.acs.org/doi/10.1021/acs.jafc.6b04815

      Helpful?

  5. Is the the standard used for estimation of cucumin/curcuminoid content in commercial turmeric samples.

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service