Skip to Content
Merck
All Photos(1)

Key Documents

M9524

Supelco

Morphine sulfate salt solution

1.0 mg/mL in methanol, drug standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H38N2O6 · H2SO4
CAS Number:
Molecular Weight:
668.75
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

drug standard

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35.CN6CC[C@]78[C@H]9Oc%10c(O)ccc(C[C@@H]6[C@@H]7C=C[C@@H]9O)c8%10

InChI

1S/2C17H19NO3.H2O4S/c2*1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;1-5(2,3)4/h2*2-5,10-11,13,16,19-20H,6-8H2,1H3;(H2,1,2,3,4)/t2*10-,11+,13-,16-,17-;/m00./s1

InChI key

USAHOPJHPJHUNS-IFCNUISUSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Guillot et al.
PloS one, 9(5), e97347-e97347 (2014-05-27)
In cats, osteoarthritis causes significant chronic pain. Chronicity of pain is associated with changes in the central nervous system related to central sensitization, which have to be quantified. Our objectives were 1) to develop a quantitative sensory testing device in
Colin Harvey-Lewis et al.
Psychopharmacology, 232(5), 885-895 (2014-09-06)
Chronic opiate use is associated with increased impulsivity in both humans and animals, and previous studies suggest that acute morphine can increase impulsivity in non-dependent rats. However, the extent to which chronic opiate usage modulates the effect of acute morphine
Laura S Stone et al.
PloS one, 9(10), e109903-e109903 (2014-10-10)
Opioids are used to manage all types of pain including acute, cancer, chronic neuropathic and inflammatory pain. Unfortunately, opioid-related adverse effects such as respiratory depression, tolerance, physical dependence and addiction have led to an underutilization of these compounds for adequate
Serotonin, morphine, and neuropathic pain: not a simple story.
Kelly Byrne et al.
Anesthesiology, 121(2), 217-218 (2014-06-03)
Nian Gong et al.
Anesthesiology, 120(4), 962-975 (2013-08-10)
D-Amino acid oxidase (DAAO) is a flavin adenine dinucleotide-dependent peroxisomal flavoenzyme which is almost exclusively expressed within astrocytes in the spinal cord. DAAO catalyzes oxidation of D-amino acids to hydrogen peroxide, which is a stable and less active reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service