Skip to Content
Merck
All Photos(1)

Key Documents

M4627

Supelco

1-Methylnicotinamide chloride

analytical standard

Synonym(s):

N-Methylnicotinic acid amide chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H9N2OCl
CAS Number:
Molecular Weight:
172.61
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5%

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

application(s)

forensics and toxicology
veterinary
vitamins, nutraceuticals, and natural products

format

neat

SMILES string

[Cl-].C[n+]1cccc(c1)C(N)=O

InChI

1S/C7H8N2O.ClH/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H

InChI key

BWVDQVQUNNBTLK-UHFFFAOYSA-N

General description

1-Methylnicotinamide (MNA) is a primary metabolite of nicotinamide produced mainly by nicotinamide N-methyltransferase (NNMT) and exhibits antithrombotic and anti-inflammatory effects.

Application

1-Methylnicotinamide chloride may be used as a standard in the quantification of 1-methylnicotinamide in urine samples using ion pairing reverse-phase high performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Urinary excretion product of niacin metabolism

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation of the niacin metabolites 1-methylnicotinamide and l-methyl-2-pyridone-5-carboxamide in random spot urine samples, by ion-pairing reverse-phase HPLC with UV detection, and the implications for the use of spot urine samples in the assessment of niacin status
Creeke IP, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 817, 247-253 (2005)
Yoshitaka Tayama et al.
Drug metabolism and pharmacokinetics, 26(1), 94-101 (2010-11-19)
Aldehyde oxidase (AO) plays an important role in metabolizing antitumor and antiviral drugs, including methotrexate, cyclophosphamide and acyclovir. Green tea and its catechins have been shown to modulate the activities of various xenobiotic-metabolizing cytochrome P450 species, both in vivo and
Jacek Miedzobrodzki et al.
Acta poloniae pharmaceutica, 67(5), 487-494 (2010-09-30)
The study was designed to demonstrate the relationship between the activity of human normal monocytes and blood platelets, to determine the metabolic activity of normal monocytes and monocytes cooperating with blood platelets in respect of their generation of reactive oxygen
Stefan Chłopicki et al.
Pharmacological reports : PR, 64(2), 369-376 (2012-06-05)
Methylnicotinamide (MNA) displays vasoprotective activity, however, the regulation of the activity of nicotinamide-N-methyltransferase (NNMT), is largely unknown. We analyze a possible involvement of IL-6 in the activation of NNMT-MNA pathway during an endurance exercise. FVB, C57Bl/6J IL6(+/+) and C57Bl/6J IL-6(-/-)
Magdalena Sternak et al.
Pharmacological reports : PR, 62(3), 483-493 (2010-07-16)
Nicotinamide N-methyltransferase (NNMT), which converts nicotinamide (NA) to 1-methylnicotinamide (MNA), is up-regulated in the cirrhotic liver. Because MNA displays PGI(2)-dependent anti-inflammatory effects, the up-regulation of NNMT may play a regulatory role in liver inflammation. In the present work, we analyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service