Skip to Content
Merck
All Photos(1)

Key Documents

M9179

Sigma-Aldrich

Meglumine

99.0-100.5% dry basis, meets USP testing specifications

Synonym(s):

N-Methyl-D-glucamine, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Beilstein:
385906
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

99.0-100.5% dry basis

form

powder

autoignition temp.

~662 °F

color

off-white

mp

129-131.5 °C (lit.)

solubility

water: soluble

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

InChI key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pentavalent Antimony Associated with G-CSF in the Treatment of Cutaneous Leishmaniasis Caused by Leishmania (Viannia) braziliensis: Investigates the use of meglumine in conjunction with G-CSF, showing potential for enhanced treatment outcomes (Suprien et al., 2024).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphaëlle Renard-Penna et al.
Radiology, 275(2), 458-468 (2015-01-20)
To compare the diagnostic performance of the magnetic resonance (MR) imaging-based Prostate Imaging Reporting and Data System (PI-RADS) and a Likert scale in the detection of prostate cancer in a cohort of patients undergoing initial prostate biopsy. This institutional review
Philipp Kickingereder et al.
Neuro-oncology, 17(8), 1139-1147 (2015-03-11)
To analyze the relevance of dynamic susceptibility-weighted contrast-enhanced MRI (DSC-MRI) derived relative cerebral blood volume (rCBV) analysis for predicting response to bevacizumab (BEV) in patients with recurrent glioblastoma (rGB). A total of 127 patients diagnosed with rGB receiving either bevacizumab
Charlotte Manisty et al.
Radiology, 276(3), 732-740 (2015-04-30)
To investigate the pharmacology and potential clinical utility of splenic switch-off to identify understress in adenosine perfusion cardiac magnetic resonance (MR) imaging. Splenic switch-off was assessed in perfusion cardiac MR examinations from 100 patients (mean age, 62 years [age range
Myriam Edjlali et al.
Radiology, 270(1), 261-268 (2013-09-14)
To assess the feasibility of a selective flow-tracking cartographic procedure applied to four-dimensional (4D) flow imaging and to demonstrate its usefulness in the characterization of dural arteriovenous fistulas (DAVFs). Institutional review board approval was obtained, and all patients provided written
Daniel M Sado et al.
Circulation. Cardiovascular imaging, 6(3), 392-398 (2013-04-09)
Anderson-Fabry disease (AFD) is a rare but underdiagnosed intracellular lipid disorder that can cause left ventricular hypertrophy (LVH). Lipid is known to shorten the magnetic resonance imaging parameter T1. We hypothesized that noncontrast T1 mapping by cardiovascular magnetic resonance would

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service