Skip to Content
Merck
All Photos(2)

Key Documents

L1886

Sigma-Aldrich

Light Green SF Yellowish

Dye content, ≥65%, certified by the Biological Stain Commission, powder

Synonym(s):

Acid Green 5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H34N2O9S3Na2
CAS Number:
Molecular Weight:
792.85
Colour Index Number:
42095
Beilstein:
5717828
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Light Green SF Yellowish, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥65%

technique(s)

titration: suitable

color

red to dark purple

mp

288 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)\C(=C3\C=CC(\C=C3)=[N+](/CC)Cc4cccc(c4)S([O-])(=O)=O)c5ccc(cc5)S([O-])(=O)=O

InChI

1S/C37H36N2O9S3.2Na/c1-3-38(25-27-7-5-9-35(23-27)50(43,44)45)32-17-11-29(12-18-32)37(31-15-21-34(22-16-31)49(40,41)42)30-13-19-33(20-14-30)39(4-2)26-28-8-6-10-36(24-28)51(46,47)48;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/p-2

InChI key

DGOBMKYRQHEFGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Light Green SF Yellowish or Acid Green 5 is a triarylmethane dye that exists as disodium salt. It has the absorption value of 629 nm.

Application

Light Green SF Yellowish functions as an efficient histological stain for collagen. It is also used as colorant for foodstuff and beverages.
Light Green SF Yellowish has been used to determine surface structures of trypsin treated tissue sections and to stain buccal cells for slide scoring.

Biochem/physiol Actions

Light Green SF Yellowish functions as an efficient histological stain for collagen. It is also used as colorant for foodstuff and beverages.

Suitability

Certified for use as a cytological counterstain and as a substitute for aniline blue in Gomori′s one-step trichrome procedure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atomic force microscope investigation of the boundary-lubricant layer in articular cartilage.
Chan SM
Osteoarthritis and Cartilage, 18, 956-956 (2010)
Buccal micronucleus cytome biomarkers may be associated with Alzheimer's disease.
Thomas P
Mutagenesis, 22, 371-371 (2007)
Decoloration treatment of a hazardous triarylmethane dye, Light Green SF (Yellowish) by waste material adsorbents
Mittal A, et al.
Journal of Colloid and Interface Science, 342(2), 518-527 (2010)
Atomic force microscope investigation of the boundary-lubricant layer in articular cartilage
Chan SMT, et al.
Osteoarthritis and Cartilage, 18(7), 956-963 (2010)
The buccal cytome and micronucleus frequency is substantially altered in Down's syndrome and normal ageing compared to young healthy controls
Thomas P, et al
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 638, 37-47 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service