Skip to Content
Merck
All Photos(4)

Key Documents

F8129

Sigma-Aldrich

Acid Fuchsin

Powder

Synonym(s):

Acid Violet 19, Fuchsin S, Fuchsin acid, Rubine S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17N3Na2O9S3
CAS Number:
Molecular Weight:
585.54
Colour Index Number:
42685
Beilstein:
4111656
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Acid Fuchsin, used in tissue staining

form

powder

composition

Dye content, ~70%

technique(s)

microbe id | staining: suitable

color

blue-green

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Cc1cc(cc(c1N)S([O-])(=O)=O)\C(c2ccc(N)c(c2)S([O-])(=O)=O)=C3/C=CC(=N)C(=C3)S(O)(=O)=O

InChI

1S/C20H19N3O9S3.2Na/c1-10-6-13(9-18(20(10)23)35(30,31)32)19(11-2-4-14(21)16(7-11)33(24,25)26)12-3-5-15(22)17(8-12)34(27,28)29;;/h2-9,23H,21-22H2,1H3,(H,24,25,26)(H,27,28,29)(H,30,31,32);;/q;2*+1/p-2

InChI key

WZRZTHMJPHPAMU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Acid Fuchsin is mainly used as a diachrome and is part of the connective tissue stains. It also works as a pH indicator.

Application

Acid Fuchsin has been used in tissue sections for Masson′s trichrome staining. It has been used as a marker to trace ingestion of solutions by green peach aphids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihiro Umebayashi et al.
Tree physiology, 27(7), 993-999 (2007-04-04)
To elucidate the water-conducting pathways in living trees by the dye injection method, suitable sample preparation procedures are needed. We evaluated quantitatively the properties and concentrations of three dyes (acid fuchsin, basic fuchsin and safranin) widely used for this purpose
Andrea Zsombok et al.
Journal of neuroscience methods, 142(1), 145-152 (2005-01-18)
By means of a condenser discharge electric shock paradigm, "dark" (compacted) granule neurons were momentarily produced in the hippocampal dentate gyri of rats, which were sacrificed either immediately or following survival periods ranging from 1h to 30 days. Except for
G Vinitha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(4), 1160-1164 (2007-08-08)
Solid-state dye-doped polymers are attractive alternative to the conventional liquid dye solutions. In this paper, nonlinear properties of the dye Acid Fuchsin has been studied. The third-order nonlinear optical properties of Acid Fuchsin dye in 1-butanol and dye-doped polymer film
Hongtao Huang et al.
Nanotechnology, 23(10), 105502-105502 (2012-03-01)
Zn-doped SnO(2) nanorods have been prepared by a simple hydrothermal method on a large scale. The as-prepared samples were characterized by x-ray powder diffraction, scanning electron microscope, transmission electron microscope, energy dispersive spectrometer, x-ray photoelectron spectroscopy, UV-vis absorption spectra and
Roberto Luongo et al.
The International journal of periodontics & restorative dentistry, 28(6), 551-557 (2009-01-17)
Platform switching is a concept recently introduced in implant dentistry. It is intended to reduce the crestal bone loss that is commonly found around implants exposed to the oral environment. The aim of this study was to examine biopsy specimens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service