Skip to Content
Merck
All Photos(1)

Documents

F0200000

Fluocinolone acetonide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Sinalar, Synandone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30F2O6
CAS Number:
Molecular Weight:
452.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluocinolone

manufacturer/tradename

EDQM

mp

267-269 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

InChI

1S/C24H30F2O6/c1-20(2)31-19-9-13-14-8-16(25)15-7-12(28)5-6-21(15,3)23(14,26)17(29)10-22(13,4)24(19,32-20)18(30)11-27/h5-7,13-14,16-17,19,27,29H,8-11H2,1-4H3/t13-,14-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

FEBLZLNTKCEFIT-VSXGLTOVSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluocinolone acetonide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soraya Rofagha et al.
Ocular immunology and inflammation, 21(1), 77-78 (2013-01-18)
To describe a case of spontaneous dissociation of a fluocinolone acetonide implant (Retisert) 5 years following implantation. Case report. A patient with chronic uveitis underwent placement of a fluocinolone acetonide implant in the right eye in 2006. The patient's inflammation
John H Kempen et al.
Ophthalmology, 120(9), 1852-1859 (2013-05-28)
To evaluate agreement between fluorescein angiography (FA) and optical coherence tomography (OCT) results for diagnosis of macular edema in patients with uveitis. Multicenter cross-sectional study. Four hundred seventy-nine eyes with uveitis from 255 patients. The macular status of dilated eyes
Bryn M Burkholder et al.
Retina (Philadelphia, Pa.), 33(8), 1684-1693 (2013-04-04)
To evaluate outcomes after placement of fluocinolone acetonide (FA) implants in eyes with birdshot chorioretinitis and to compare these outcomes with eyes with posterior and panuveitis. This is a retrospective cohort study of 48 eyes from patients with posterior and
Raymond Iezzi et al.
Biomaterials, 33(3), 979-988 (2011-11-04)
Retinal neuroinflammation, mediated by activated microglia, plays a key role in the pathogenesis of photoreceptor and retinal pigment epithelial cell loss in age-related macular degeneration and retinitis pigmentosa. Targeted drug therapy for attenuation of neuroinflammation in the retina was explored
Benjamin P Nicholson et al.
American journal of ophthalmology, 154(6), 969-973 (2012-09-18)
To characterize the phenomenon of dissociation of the 2 components of the sustained-release fluocinolone acetonide implant (Retisert; Bausch & Lomb) during removal or exchange procedures, or both, and to evaluate outcomes after these events. Retrospective, observational case series. Retrospective review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service