Skip to Content
Merck
All Photos(1)

Key Documents

E0085000

Eicosapentaenoic acid ethyl ester

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

eicosapentaenoic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC)C(=O)\C=C\C=C\C=C\C=C\C=C\CCCCCCCCC

InChI

1S/C22H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h12-21H,3-11H2,1-2H3/b13-12+,15-14+,17-16+,19-18+,21-20+

InChI key

DTEMJWLYSQBXEL-MBFZXKRTSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Eicosapentaenoic acid ethyl ester EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Young Park et al.
Journal of agricultural and food chemistry, 53(21), 8334-8341 (2005-10-13)
The effects of the addition of amino acids and peptides on the oxidation of eicosapentaenoic acid ethyl ester (EPE) encapsulated by maltodextrin (MD) were investigated. The encapsulated lipid was prepared in two steps, that is, by mixing of EPE with
Christie M Ballantyne et al.
The American journal of cardiology, 110(7), 984-992 (2012-07-24)
AMR101 is an ω-3 fatty acid agent containing ≥96% pure icosapent-ethyl, the ethyl ester of eicosapentaenoic acid. The efficacy and safety of AMR101 were evaluated in this phase 3, multicenter, placebo-controlled, randomized, double-blinded, 12-week clinical trial (ANCHOR) in high-risk statin-treated
Abbie L Cawood et al.
Atherosclerosis, 212(1), 252-259 (2010-06-15)
To examine n-3 polyunsaturated fatty acid (PUFA) incorporation into atherosclerotic plaques and the association with plaque inflammation and stability. Patients awaiting carotid endarterectomy (n=121) were randomised to consume control capsules or n-3 PUFA ethyl ester capsules until surgery (median 21
Yoshiyuki Kimura et al.
Cancer science, 96(7), 441-450 (2005-08-02)
The aim of the present work was to compare the effects of eicosapentaenoic acid (EPA) ethylester and EPA derivatives (EPAD) on tumor growth and metastasis to the lung in tumor-bearing mice. Because EPA is very unstable during long-term storage, the
Terry A Jacobson
Expert review of cardiovascular therapy, 10(6), 687-695 (2012-08-17)
ω-3 fatty acids reduce triglyceride (TG) levels, but corresponding increases in low-density lipoprotein cholesterol (LDL-C) levels may compromise achievement of lipid goals in patients with elevated cardiovascular risk. AMR101 is an investigational agent containing ≥96% of pure icosapent ethyl (the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service