Skip to Content
Merck
All Photos(2)

Key Documents

D2410

Sigma-Aldrich

cis-4,7,10,13,16,19-Docosahexaenoic acid ethyl ester

≥97%

Synonym(s):

DHA ethyl ester, Ethyl 4,7,10,13,16,19-docosahexaenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O2
CAS Number:
Molecular Weight:
356.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

algae

Assay

≥97%

form

liquid

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCOC(=O)CC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC

InChI

1S/C24H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-4-2/h5-6,8-9,11-12,14-15,17-18,20-21H,3-4,7,10,13,16,19,22-23H2,1-2H3/b6-5+,9-8+,12-11+,15-14+,18-17+,21-20+

InChI key

ITNKVODZACVXDS-YATCGRJWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidefumi Yoshii et al.
Bioscience, biotechnology, and biochemistry, 66(4), 749-753 (2002-05-31)
The application of omega-3 polyunsaturated fatty acids (PUFAs) as food additives is restricted by their chemically quite reactive properties. However, quantitative analyses of the oxidative kinetics of PUFAs are very few compared to other studies on food chemistry. In this
I Ozawa et al.
Biotechnology progress, 17(5), 893-896 (2001-10-06)
Docosahexaenoic acid ethyl ester (DHA-Et) was purified by adsorption on Ag-ion-immobilized membranes via selective interaction between silver ion and carbon-carbon double bonds of DHA-Et. Silver ions were immobilized onto sulfonic-acid-group-containing porous hollow-fiber membranes at an Ag ion density of 1.4
R C Poulsen et al.
Calcified tissue international, 81(6), 459-471 (2007-11-17)
Long chain polyunsaturated fatty acids (LCPUFAs) are involved in the regulation of bone metabolism. Increased dietary consumption of n-3, and possibly some n-6, LCPUFAs may limit postmenopausal bone loss. The aim of this study was to determine the effects on
Yoshiyuki Murata et al.
Bioscience, biotechnology, and biochemistry, 68(3), 743-745 (2004-04-02)
Ethyl 4,7,10,13,16,19-docosahexaenoate (DHA-Et) dissolved in an ethanol solution or embedded in liposomes was degraded by irradiating with gamma rays in a dose-dependent manner. The degradation rate of DHA-Et embedded in liposomes was higher than that of DHA-Et dissolved in ethanol.
Hideaki Kabuto et al.
Neurochemical research, 34(7), 1299-1303 (2009-02-17)
Superoxide and hydroxyl radicals are implicated in the pathogenesis of Parkinson disease, and induction of lipid peroxidation is an important factor in progression of this disease. Docosahexaenoic acid (DHA) is a key component of the cell membrane, and its peroxidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service