Skip to Content
Merck
All Photos(2)

Key Documents

53171

Supelco

cis-4,7,10,13,16,19-Docosahexaenoic acid

analytical standard

Synonym(s):

DHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Beilstein:
1715505
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(O)=O

InChI

1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-

InChI key

MBMBGCFOFBJSGT-KUBAVDMBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinglong Chen et al.
International journal of molecular sciences, 22(11) (2021-06-03)
Oxidative stress occurs in a variety of clinical liver diseases and causes cellular damage and mitochondrial dysfunction. The clearance of damaged mitochondria by mitophagy may facilitate mitochondrial biogenesis and enhance cell survival. Although the supplementation of docosahexaenoic acid (DHA) has
Physiology: Double function at the blood-brain barrier.
Christer Betsholtz
Nature, 509(7501), 432-433 (2014-05-16)
Lucy M Browning et al.
The Journal of nutrition, 144(5), 667-672 (2014-03-22)
Consumption of oily fish is sporadic, whereas controlled intervention studies of n-3 (ω-3) fatty acids usually provide capsules containing eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) as a daily dose. This methodologic study explored whether there are differences in the
Y Freund Levi et al.
Journal of internal medicine, 275(4), 428-436 (2014-01-15)
Little is known about the transfer of essential fatty acids (FAs) across the human blood-brain barrier (BBB) in adulthood. In this study, we investigated whether oral supplementation with omega-3 (n-3) FAs would change the FA profile of the cerebrospinal fluid
Four-year follow-up of children born to women in a randomized trial of prenatal DHA supplementation.
Maria Makrides et al.
JAMA, 311(17), 1802-1804 (2014-05-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service