Skip to Content
Merck
All Photos(3)

Key Documents

D172405

Sigma-Aldrich

N,N-Dimethyl-4-nitrosoaniline

97%, powder or chunks

Synonym(s):

4-Nitroso-N,N-dimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ONC6H4N(CH3)2
CAS Number:
Molecular Weight:
150.18
Beilstein:
607293
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

N,N-Dimethyl-4-nitrosoaniline, ≥97%

Quality Level

Assay

≥97%

form

powder or chunks

mp

85-87 °C (lit.)

solubility

ethanol: 5% (green to very dark green)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)N=O

InChI

1S/C8H10N2O/c1-10(2)8-5-3-7(9-11)4-6-8/h3-6H,1-2H3

InChI key

CMEWLCATCRTSGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethyl-4-nitrosoaniline has been used along with imidazole to detect the formation of singlet oxygen in cells.

Biochem/physiol Actions

N,N-Dimethyl-4-nitrosoaniline is used for the detection of singlet oxygen. Singlet oxygen reacts with imidazole and thereby bleaches N,N-Dimethyl-4-nitrosoaniline via oxidation. Bleaching of N,N-Dimethyl-4-nitrosoaniline is observed as reduction in the absorption at 438nm.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Self-heat. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Marie Schreiber et al.
Environmental science & technology, 39(10), 3811-3818 (2005-06-15)
The formation of the potent carcinogen, N-nitrosodimethylamine (NDMA), during chlorine disinfection has caused significant concern among drinking water and wastewater recycling utilities practicing intentional or unintentional chloramination. Previous research modeled NDMA formation as arising from a reaction between monochloramine and
S Kapur et al.
Molecular psychiatry, 7(8), 837-844 (2002-09-17)
Ketamine and PCP are commonly used as selective NMDA receptor antagonists to model the putative hypoglutamate state of schizophrenia and to test new antipsychotics. Recent findings question the NMDA receptor selectivity of these agents. To examine this further, we measured
Elif Pehlivanoglu-Mantas et al.
Water research, 40(6), 1287-1293 (2006-03-01)
To assess the stability of precursors of the chloramine disinfection byproduct N-nitrosodimethylamine (NDMA) under conditions expected in effluent-dominated surface waters, effluent samples from four municipal wastewater treatment plants were subjected to chlorination and chloramination followed by incubation in the presence
Juliette Gafni et al.
Toxicological sciences : an official journal of the Society of Toxicology, 77(1), 72-82 (2003-11-06)
The mechanisms by which non-coplanar 2,2',3,5',6-pentachlorobiphenyl (PCB 95) and rapamycin interact with ryanodine receptor (RyR) complexes to alter Ca2+ signaling, were explored in intact cerebellar granule neurons. PCB 95 (10 microM, 20 min) significantly increased the number of neurons responding
Zhuo Chen et al.
Environmental science & technology, 40(23), 7290-7297 (2006-12-22)
This paper presents mechanistic studies on the formation of NDMA, a newly identified chloramination disinfection byproduct, from reactions of monochloramine with natural organic matter. A kinetic model was developed to validate proposed reactions and to predict NDMA formation in chloraminated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service