Skip to Content
Merck
All Photos(1)

Documents

CLS431752

Corning® cell strainer

pore size 100 μm, yellow, sterile, pkg of (individually wrapped), pack of 50 ea

Synonym(s):

Cell strainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104932
NACRES:
NB.24

material

Nylon
yellow

sterility

sterile

packaging

pack of 50 ea
pkg of (individually wrapped)

manufacturer/tradename

Corning 431752

pore size

100 μm

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 100µm Cell Strainer is a sterile, easy-to-use device for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Applications include stem cells and primary cells. Individually packaged, 50/case.

Application

  • Faster, easier alternative to gauze filtration
  • Ideal for preparation of flow cytometry samples, including:
    Single cell suspensions of blood cells from marrow, pancreas, thymus, tonsil, and lymph nodes
    Stem cells, tissue-derived cells, and cancer cells
    Preparation of specimens for primary cell cultures and immunogens
    Preparation of freezing stocks
    Filtering agglutinative proteins produced in inactivation serum

Features and Benefits

  • Strong nylon mesh with 100- micron pores for optimal performance in a variety of applications
  • Evenly spaced mesh pores providing consistent and reliable results
  • Conveniently accessible in individual packaging
  • Extended lip on strainer enables aseptic handling with forceps
  • Molded color-coded polypropylene frame with tab enables easy handling and identification
  • Fits all major brands of 50mL Conical Tube
  • Disposable, easy-to-use, inexpensive, maintains sample integrity
  • Sterilized by gamma irradiation, noncytotoxic

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arjun Challagulla et al.
Microorganisms, 9(1) (2021-01-17)
Marek's disease (MD), caused by MD herpesvirus (MDV), is an economically important disease in chickens. The efficacy of the existing vaccines against evolving virulent stains may become limited and necessitates the development of novel antiviral strategies to protect poultry from
Emma Lång et al.
Nature communications, 9(1), 3665-3665 (2018-09-12)
Epithelial sheet spreading is a fundamental cellular process that must be coordinated with cell division and differentiation to restore tissue integrity. Here we use consecutive serum deprivation and re-stimulation to reconstruct biphasic collective migration and proliferation in cultured sheets of
Zhi Chen et al.
American journal of translational research, 12(6), 2379-2395 (2020-07-14)
Inflammation is a leading cause of severe acute pancreatitis (SAP). MicroRNAs (miRNAs) are emerging as important regulators involved in the pathogenesis of many diseases including pancreatitis. To identify miRNAs that contribute to the pathology of SAP, we carried out a
Joanna Kalucka et al.
Cell, 180(4), 764-779 (2020-02-16)
The heterogeneity of endothelial cells (ECs) across tissues remains incompletely inventoried. We constructed an atlas of >32,000 single-EC transcriptomes from 11 mouse tissues and identified 78 EC subclusters, including Aqp7+ intestinal capillaries and angiogenic ECs in healthy tissues. ECs from
Christinna V Jørgensen et al.
Journal of pharmacological and toxicological methods, 97, 59-66 (2019-04-05)
Adipose tissues release adipokines, which regulate energy intake and expenditure. G protein-coupled receptors (GPCRs) and associated signaling pathways in adipocytes are potentially important drug targets for conditions with disturbed energy metabolism. The aim of the current study was to compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service