Skip to Content
Merck
All Photos(3)

Key Documents

272442

Sigma-Aldrich

Reichardt′s dye

Dye content 90 %

Synonym(s):

2,6-Diphenyl-4-(2,4,6-triphenyl-1-pyridinio)phenolate, 2,6-Diphenyl-4-(2,4,6-triphenylpyridinio)phenolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H29NO
CAS Number:
Molecular Weight:
551.68
Beilstein:
1444580
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 90%

technique(s)

titration: suitable

mp

271-275 °C (lit.)

λmax

306 nm
551 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-]c1c(cc(cc1-c2ccccc2)-[n+]3c(cc(cc3-c4ccccc4)-c5ccccc5)-c6ccccc6)-c7ccccc7

InChI

1S/C41H29NO/c43-41-37(31-18-8-2-9-19-31)28-36(29-38(41)32-20-10-3-11-21-32)42-39(33-22-12-4-13-23-33)26-35(30-16-6-1-7-17-30)27-40(42)34-24-14-5-15-25-34/h1-29H

InChI key

UWOVWIIOKHRNKU-UHFFFAOYSA-N

General description

Reichardt′s dye belongs to the class of azo merocyanine dyes. Reichardt′s dye, also known as ET(30) or pyridinium N-phenoxyde betaine dye is a highly solvatochromic betaine dye useful for determining solvent polarity effects. It plays an important role as a probe of solvent polarity in biological systems and on surfaces.

Application

Reichardt’s dye, a solvatochromic betaine dye has been used for the determination of solvent polarity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical Reviews, 94, 2319-2319 (1994)
Beatrice Swensson et al.
Polymers, 12(6) (2020-06-12)
To further understand cellulose-solvent interactions in aqueous hydroxide solutions, cellulose behavior in aqueous solutions of NaOH combined with tetramethylammonium hydroxide (TMAH) or benzyltrimethylammonium hydroxide (Triton B), as well as urea, was investigated. The rheological properties of the solutions were assessed
C. Machado et al.
Journal of Chemical Education, 78, 649-649 (2001)
Yu Chen et al.
Analytica chimica acta, 1034, 128-136 (2018-09-09)
In this work, a paper-based optoelectronic sensor (paper-nose) is presented for sensing volatile gases in air. The proposed optoelectronic sensor is a combination of both colorimetric (optical) and chemiresistive (electronic) sensor arrays in order to improve the selectivity of the
C. Reichardt
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 80, 1415-1415 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service