Skip to Content
Merck
All Photos(1)

Key Documents

C1359

Sigma-Aldrich

Calcein AM solution

≥90% purity (HPLC), solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H46N2O23
CAS Number:
Molecular Weight:
994.86
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution

Quality Level

Assay

≥90% (HPLC)

form

solution

concentration

4 mM in DMSO

technique(s)

titration: suitable

color

colorless
white to off-white

λmax

491-497 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)Cc1c(OC(C)=O)ccc2c1Oc3c(CN(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)c(OC(C)=O)ccc3C24OC(=O)c5ccccc45

InChI

1S/C46H46N2O23/c1-25(49)60-21-64-39(55)17-47(18-40(56)65-22-61-26(2)50)15-32-37(68-29(5)53)13-11-35-43(32)70-44-33(16-48(19-41(57)66-23-62-27(3)51)20-42(58)67-24-63-28(4)52)38(69-30(6)54)14-12-36(44)46(35)34-10-8-7-9-31(34)45(59)71-46/h7-14H,15-24H2,1-6H3

InChI key

XKFSBWQWNMZWFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

In cells, calcein AM solution has been used in calcein-AM dye transfer assay. It has been used for the staining of cells to study quantification of cell-migration. It has been used to study cytotoxic effects of drugs in cells.

Biochem/physiol Actions

Calcein AM is a membrane permeable nonfluorescent molecule. It is converted to fluorescent calcein by intracellular esterases. Calcein has limited membrane permeability. Calcein AM is also a substrate for p-glycoprotein (Pgp). Presence of Pgp results in efflux of calcein AM and thereby decreases cellular fluorescence. On the other hand, presence of Pgp inhibitor increases cellular entry of calcein AM and thereby increases cellular fluorescence. Calcein AM plays an important role in early drug discovery.
Fluorescent cell permeable derivative of calcein.

Physical form

Solution in 0.4% Calcein AM and 99.6% DMSO.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of connexin 43 reduces melanoma proliferative and metastatic capacity.
Tittarelli A, Guerrero I, et al.
British Journal of Cancer, 115, e14-e14 (2016)
Pharmaceutical Profiling in Drug Discovery for Lead Selection (2005)
PEA15 impairs cell migration and correlates with clinical features predicting good prognosis in neuroblastoma.
Gawecka JE, et al.
International Journal of Cancer. Journal International Du Cancer, 131, 1556-1556 (2012)
Xiao-Cong Huang et al.
Biochemical pharmacology, 85(9), 1257-1268 (2013-02-19)
High intrinsic or acquired expression of membrane spanning, adenosine triphosphate binding cassette (ABC) transporter proteins, such as P-glycoprotein (P-gp), in cancers represents a major impediment to chemotherapy, with accelerated drug efflux leading to multi-drug resistance (MDR). Although ABC transporter inhibitors
Bao-Ling Du et al.
Journal of biomedical materials research. Part A, 103(4), 1533-1545 (2014-07-22)
Biological materials combined with genetically-modified neural stem cells (NSCs) are candidate therapy targeting spinal cord injury (SCI). Based on our previous studies, here we performed gelatin sponge (GS) scaffold seeded with neurotrophin-3 (NT-3) and its receptor TrkC gene modifying NSCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service