Skip to Content
Merck
All Photos(1)

Documents

CBA415

Sigma-Aldrich

Live Dead Cell Viability Assay Kit for 3D and 2D cell culture

Live-dead cell viability kit for 3D and 2D cell cultures. Indicates live, dead and total cell count. Sufficient for 5 x 24-well plates or 12 x 96-well plates.

Synonym(s):

Viability Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

detection method

fluorometric

General description

The Live-Dead Cell Viability Assay Kit is a quick and simple three-color assay to measure cell viability. The kit consists of Calcein-AM (stains live cells), Propidium Iodide (stains dead cells) and Hoechst 33342 (stains all cells). The kit has been optimized for 3D cell culture (spheroids, human organoids and 3D matrices) and 2D cell culture (multiple cell types). The kit can be used for flow cytometry, fluorescence microscopy and with fluorescence microplate readers. The kit determines viability of cells based on plasma membrane integrity and esterase activity.

One kit contains reagents sufficient for 5 x 24-well plates or 12 x 96-well plates.

<bold>Advantages:</bold>
1. Tested and optimized for 3D cell culture, Spheroid culture, organoid culture, 2D cell culture and flow cytometry.
2. Easy to use and quick protocol.
3. Stronger intensity of Calcein AM and Hoechst dyes compared to other products.

Spectral Properties
Calcein AM: Ex/Em: 490/515 nm (Green)
Propidium Iodide: Ex/Em: 535/617 nm (Red)
Hoechst 33342: Ex/Em: 361/486 nm (Blue)

Protocols:
Protocols for staining 2D monolayer cell culture, 3D cell culture, 3D spheroids and 3D organoids are included in the datasheet.

Storage and Stability

Store at -20oC or below, and protect from light. The vials can be frozen and thawed upto 10 times. Before use, allow the vials to thaw to room temperature and centrifuge briefly.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service