Skip to Content
Merck
All Photos(1)

Key Documents

BP578

Dexamethasone

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dexamethasone

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

262-264 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Tobramycin and Dexamethasone Eye Drops, Suspension
  • Dexamethasone Sodium Phosphate Oral Solution
  • Hydrocortisone Sodium Succinate Injection
  • Prednisolone Tablets
  • Dexamethasone Eye Drops, Suspension
  • Dexamethasone Sodium Phosphate Eye Drops, Solution
  • Dexamethasone and Neomycin Ear Spray
  • Betamethasone Tablets
  • Dexamethasone Tablets
  • Biochem/physiol Actions

    Glucocorticoid anti-inflammatory agent
    Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Health hazard

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Repr. 1B

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Alexandre Denoyer et al.
    Ophthalmology, 122(4), 669-676 (2014-12-03)
    To compare small incision lenticule extraction (SMILE) versus LASIK for post-refractive dry eye disease. Prospective, comparative, nonrandomized clinical study. Thirty patients scheduled for bilateral myopic SMILE and 30 age-, sex-, and refraction-matched patients scheduled for bilateral myopic LASIK were enrolled
    Michael J White et al.
    Cell, 159(7), 1549-1562 (2014-12-20)
    Activated caspases are a hallmark of apoptosis induced by the intrinsic pathway, but they are dispensable for cell death and the apoptotic clearance of cells in vivo. This has led to the suggestion that caspases are activated not just to kill
    A Keith Stewart et al.
    The New England journal of medicine, 372(2), 142-152 (2014-12-09)
    Lenalidomide plus dexamethasone is a reference treatment for relapsed multiple myeloma. The combination of the proteasome inhibitor carfilzomib with lenalidomide and dexamethasone has shown efficacy in a phase 1 and 2 study in relapsed multiple myeloma. We randomly assigned 792
    Sharon D'Souza et al.
    Investigative ophthalmology & visual science, 55(6), 3396-3402 (2014-05-08)
    New femtosecond laser platforms may reduce ocular surface interference and LASIK-associated dry eye. This study investigated tear protein profiles in subjects who underwent LASIK using two femtosecond lasers to assess differences in protein expression. This was a randomized interventional clinical
    D Rhebergen et al.
    Psychoneuroendocrinology, 51, 341-350 (2014-12-03)
    Altered functioning of the hypothalamic-pituitary-adrenal axis (HPA-axis) has been associated with depression, but findings have been inconsistent. Among older depressed persons, both hyperactivity and hypo-activity of the HPA-axis were demonstrated. However, most studies were population-based studies, with single cortisol measurements

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service