Skip to Content
Merck
All Photos(1)

Key Documents

BCR091

Anthanthrene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
2052392
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1cc2ccc3cc4cccc5ccc6cc(c1)c2c3c6c45

InChI

1S/C22H12/c1-3-13-7-9-18-12-16-6-2-4-14-8-10-17-11-15(5-1)19(13)21(18)22(17)20(14)16/h1-12H

InChI key

YFIJJNAKSZUOLT-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR091

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiqiang Zhao et al.
Science advances, 6(22), eaba6714-eaba6714 (2020-06-12)
Two-dimensional van der Waals heterojunctions (2D-vdWHs) stacked from atomically thick 2D materials are predicted to be a diverse class of electronic materials with unique electronic properties. These properties can be further tuned by sandwiching monolayers of planar organic molecules between
[Accelerated development of an induced soft tissue sarcoma in hypokinesia].
M A Makarov et al.
Voprosy onkologii, 29(2), 96-99 (1983-01-01)
E L Cavalieri et al.
Journal of cancer research and clinical oncology, 115(1), 67-72 (1989-01-01)
Comparative studies of tumor-initiating activity in mouse skin and carcinogenicity in rat mammary gland were conducted with several dibenzo[a]-pyrenes (DBPs). SENCAR mice were initiated with DB[a, e]P, DB[a, h]P, DB[a, i]P, DB[a, l]P and anthanthrene, and promoted with tetradecanoyl-phorbol acetate.
Partha Sarathi Mukherjee et al.
Journal of the American Chemical Society, 126(8), 2464-2473 (2004-02-26)
A series of neutral, platinum-based macrocycles was synthesized from rigid oxygen donor building blocks via self-assembly. The combination of a platinum-based 60 degrees acceptor unit 1 with several linear and angular dicarboxylate bridging ligands afforded hitherto unknown neutral platinum-based supramolecular
Baiping Han et al.
The Journal of chemical physics, 130(24), 244502-244502 (2009-07-02)
In this paper, we investigate the properties of photon emission statistics of single molecule in solid matrix. The influences of solid matrix surroundings on photon emission of single molecule system under the laser field and rf field for several examples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service