Skip to Content
Merck
All Photos(1)

Documents

804541

Sigma-Aldrich

Quaterrylene

Synonym(s):

Benzo[1,2,3-cd: 4,5,6-c′d′]diperylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H20
CAS Number:
Molecular Weight:
500.59
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

density

1.467±0.06 g/cm3 (lit.)

SMILES string

C12=CC=CC(C3=C(C4=CC=C5C6=CC=C(C7=CC=C8)C9=C(C%10=C7C8=CC=C%10)C=CC%11C96)C5=C%11C=C3)=C1C4=CC=C2

InChI

1S/C40H22/c1-5-21-6-2-10-24-28-14-18-32-34-20-16-30-26-12-4-8-22-7-3-11-25(36(22)26)29-15-19-33(40(34)38(29)30)31-17-13-27(37(28)39(31)32)23(9-1)35(21)24/h1-20,31,39H

InChI key

BWASZFNXSFVOCT-UHFFFAOYSA-N

Application

A promising candidate for organic electronic applications because it absorbs a broad spectrum of UV and visible light, while possessing excellent thermal and oxidative stability and a low HOMO-LUMO band gap.
  • Organic Electronics
  • Photovoltaic Components
  • Graphene Nanoribbons
  • Electroactive Polymers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh Thamatam et al.
Chemical communications (Cambridge, England), 49(80), 9122-9124 (2013-09-04)
Quaterrylene is prepared in a single reaction and high yield by Scholl-type coupling of perylene, utilizing trifluoromethanesulfonic acid as catalyst and DDQ or molecular oxygen as oxidant. Dissolution in 1 M triflic acid/dichloroethane with sonication yields the aromatic quaterrylene oxidative
Synthesis and Photophysics of Quaterrylene Molecules in Single-Walled Carbon Nanotubes: Excitation Energy Transfer between a Nanoscale Cylinder and Encapsulated Molecules
Koyama et al.
The Journal of Physical Chemistry C, 118(37), 21671-21681 (2014)
Ambipolar carrier transport in hetero-layered organic transistors consisting of quaterrylene and N,N'-dioctyl-3,4,9,10-perylenedicarboximide
Hiroshiba et al.
Organic Electronics, 12(8), 1336-1340 (2011)
On the Way to Graphene: The Bottom-Up Approach to Very Large PAHs Using the Scholl Reaction
Cataldo et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 19(8), 713-725 (2011)
Exciton dynamics at the heteromolecular interface between N,N'-dioctyl-3,4,9,10-perylenedicarboximide and quaterrylene, studied using time-resolved photoluminescence
Hiroshiba et al.
AIP Advances, 4(6), 067112/1-067112/11 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service