Skip to Content
Merck
All Photos(1)

Documents

60615

Sigma-Aldrich

Kanamycin sulfate

mixture of Kanamycin A (main component) and Kanamycin B and C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H36-37N4-5O10-11 · H2SO4
CAS Number:
Beilstein:
3874279
MDL number:
UNSPSC Code:
51281654
PubChem Substance ID:
NACRES:
NA.85

Quality Level

form

powder or crystals

potency

≥750 units per mg (calculated with reference to dried substance)

ign. residue

≤0.5% (as SO4)

loss

≤1.5% loss on drying

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

protein synthesis | interferes

SMILES string

OS(O)(=O)=O.NC[C@H]1O[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

General description

Chemical structure: aminoglycoside

Application

Kanamycin sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium.†† Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100 μg/mL.

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance: Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial Spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Packaging

5g, 25 g

Caution

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.

Preparation Note

Kanamycin sulfate is soluble in water at 50 mg/mL, yielding a clear solution. It is practically insoluble in alcohol, acetone, chloroform, ether and ethyl acetate. A 1% solution in water has a pH of 6.5 to 8.5. Sterile solutions can be prepared by a sterile filtration, through a .2μm filter.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.Storage class (TRGS 510): Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 7718-7732 (2020-04-16)
Liver inflammation is a common extraintestinal manifestation in inflammatory bowel disease (IBD), yet, the mechanisms driving gut-liver axis inflammation remain poorly understood. IBD leads to a breakdown in the integrity of the intestinal barrier causing an increase in portal and
Lyly Luhachack et al.
Nature communications, 10(1), 2868-2868 (2019-06-30)
Prokaryotes and eukaryotes alike endogenously generate the gaseous molecule hydrogen sulfide (H2S). Bacterial H2S acts as a cytoprotectant against antibiotics-induced stress and promotes redox homeostasis. In E. coli, endogenous H2S production is primarily dependent on 3-mercaptopyruvate sulfurtransferase (3MST), encoded by
Jia Zheng et al.
Science (New York, N.Y.), 365(6451), 347-353 (2019-07-28)
Cryptic genetic variation can facilitate adaptation in evolving populations. To elucidate the underlying genetic mechanisms, we used directed evolution in Escherichia coli to accumulate variation in populations of yellow fluorescent proteins and then evolved these proteins toward the new phenotype
Sristy Shikha et al.
Scientific reports, 10(1), 1463-1463 (2020-01-31)
Microbes develop several strategies to survive in the adverse condition such as biofilm formation, attaining non-dividing state, altering drug target or drug, thereby increases the burden of drug dosage. To combat these issues, nanoparticles have shown an alternative approach for
Evangelia Vogiatzaki et al.
Current biology : CB, 27(19), 2893-2900 (2017-09-26)
Seed production requires the transfer of nutrients from the maternal seed coat to the filial endosperm and embryo. Because seed coat and filial tissues are symplasmically isolated, nutrients arriving in the seed coat via the phloem must be exported to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service