Skip to Content
Merck
All Photos(1)

Key Documents

93598

Supelco

Itaconic acid

analytical standard

Synonym(s):

Methylenesuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C(=CH2)CO2H
CAS Number:
Molecular Weight:
130.10
Beilstein:
1759501
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

autoignition temp.

1472 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

165-168 °C (lit.)

density

1.573 g/mL at 25 °C (lit.)

format

neat

SMILES string

OC(=O)CC(=C)C(O)=O

InChI

1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h1-2H2,(H,6,7)(H,8,9)

InChI key

LVHBHZANLOWSRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Itaconic acid, an unsaturated dicarbonic organic acid, is a metabolic product of Aspergillus itaconicus.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Klement et al.
Bioresource technology, 135, 422-431 (2013-01-10)
In the last years, itaconic acid has gained increasing interest as future bio-based platform chemical. To replace petrol-based compounds such as methacrylic acid in industry, the economic efficiency of the current biotechnological production processes with the fungus Aspergillus terreus has
Jahminy Sasikaran et al.
Nature chemical biology, 10(5), 371-377 (2014-03-25)
Itaconate (methylenesuccinate) was recently identified as a mammalian metabolite whose production is substantially induced during macrophage activation. This compound is a potent inhibitor of isocitrate lyase, a key enzyme of the glyoxylate cycle, which is a pathway required for the
Sambamurthy K.
Pharmaceutical Biotechnology, 56(3-4), 289-295 (2006)
Metabolism: pathogens love the poison.
Stéphane Ménage et al.
Nature chemical biology, 10(5), 326-327 (2014-04-20)
T Willke et al.
Applied microbiology and biotechnology, 56(3-4), 289-295 (2001-09-11)
Itaconic acid (IA) is an unsaturated dicarbonic organic acid. It can easily be incorporated into polymers and may serve as a substitute for petrochemical-based acrylic or methacrylic acid. It is used at 1-5% as a co-monomer in resins and also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service