Skip to Content
Merck
All Photos(1)

Key Documents

122750

Sigma-Aldrich

trans-Aconitic acid

98%

Synonym(s):

trans-Propene-1,2,3-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
174.11
Beilstein:
1725830
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

190 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C\C(=C/C(O)=O)C(O)=O

InChI

1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1+

InChI key

GTZCVFVGUGFEME-HNQUOIGGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-Aconitic acid is inhibitor of the tricarboxylic acid cycle and was identified in range grasses.

trans-Aconitic acid is an antioxidant used in food and cosmetics preparation and also used as a plasticizer in the polymer industry.

Application

trans-Aconitic acid was used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.

Biochem/physiol Actions

Antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitation of trans-aconitic acid in different stages of the sugar-manufacturing process
Montoya, Guillermo and Londono, et al.
Journal of Agricultural and Food Chemistry, 62, 8314-8831 (2014)
Effect of catalytic conditions on the synthesis of new aconitate esters.
Piang-Siong W, et al.
Industrial Crops and Products, 35(1), 203-210 (2012)
R Burau et al.
Science (New York, N.Y.), 150(3697), 766-767 (1965-11-05)
trans-Aconitate ion, an inhibitor of the tricarboxylic acid cycle, was identified in range grasses as trans-aconitic acid, which was isolated in crystalline form. It occurs in surprisingly high concentrations in early-season forage grasses. Dry-weight concentrations of trans-aconitate vary with season
Contribution of trans-aconitic acid to DPPH scavenging ability in different media
Piang-Siong, William and De Caro, et al
Food Chemistry, 214, 447-452 (2017)
Effect of Cross-Linking Agent Chemistry and Coating Conditions on Physical, Chemical and Separation Properties of PVA-PSf Composite Membranes.
Dlamini DS, et al. et al.
Separation Science and Technology (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service