Skip to Content
Merck
All Photos(4)

Key Documents

A3412

Sigma-Aldrich

cis-Aconitic acid

≥98%

Synonym(s):

cis-Propene-1,2,3-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
174.11
Beilstein:
1725829
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
≥98%

Quality Level

Assay

≥98%

form

powder

mp

122 °C (lit.)

storage temp.

−20°C

SMILES string

OC(=O)C\C(=C\C(O)=O)C(O)=O

InChI

1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1-

InChI key

GTZCVFVGUGFEME-IWQZZHSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-Aconitic acid is an organic acid mainly found in plants. It is one of the intermediates formed during citric acid cycle.

Application

cis-Aconitic acid can be used as a standard for the determination of organic acids in plant tissues.

Caution

May form trans isomer upon prolonged storage

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Federico Rojas et al.
PLoS neglected tropical diseases, 15(4), e0009284-e0009284 (2021-04-29)
The ability to reproduce the developmental events of trypanosomes that occur in their mammalian host in vitro offers significant potential to assist in understanding of the underlying biology of the process. For example, the transition from bloodstream slender to bloodstream
Esterification and gas chromatography of some acids of the tricarboxylic acid cycle.
McKeown G G and Read S I
Analytical Chemistry, 37(13), 1780-1781 (1965)
Ramji Rathod et al.
Metabolites, 10(3) (2020-03-18)
The tricarboxylic acid (TCA) cycle is a central part of carbon and energy metabolism, also connecting to glycolysis, amino acid, and lipid metabolism. The quantitation of the TCA cycle intermediate within one method is lucrative due to the interest in
Determination of organic acids in tissues and exudates of maize, lupin, and chickpea by high-performance liquid chromatography? tandem mass spectrometry.
Erro J, et al.
Journal of Agricultural and Food Chemistry, 57(10), 4004-4010 (2009)
Junyoung Kim et al.
Scientific reports, 7, 39768-39768 (2017-01-05)
Itaconate, a C5 unsaturated dicarboxylic acid, is an important chemical building block that is used in manufacturing high-value products, such as latex and superabsorbent polymers. Itaconate is produced by fermentation of sugars by the filamentous fungus Aspergillus terreus. However, fermentation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service