Skip to Content
Merck
All Photos(1)

Key Documents

A8346

Sigma-Aldrich

ProteoMass ACTH Fragment 18-39 MALDI-MS Standard

vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da

Synonym(s):

Adrenocorticotropic hormone fragment 18-39

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

mol wt

monoisotopic mol wt 2,464.1989 Da

Quality Level

packaging

vial of 10 nmol

SMILES string

CC(C)CC(NC(=O)C1CCCN1C(=O)C(Cc2ccccc2)NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(C)NC(=O)C(CO)NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(C)NC(=O)CNC(=O)C(CC(N)=O)NC(=O)C3CCCN3C(=O)C(Cc4ccc(O)cc4)NC(=O)C(NC(=O)C(CCCCN)NC(=O)C(NC(=O)C5CCCN5C(=O)C(N)CCCNC(N)=N)C(C)C)C(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(Cc6ccccc6)C(O)=O

InChI

1S/C112H165N27O36/c1-56(2)48-72(100(163)125-70(37-41-86(148)149)97(160)133-77(111(174)175)51-63-24-14-11-15-25-63)129-103(166)79-28-20-46-138(79)109(172)75(49-62-22-12-10-13-23-62)131-93(156)61(9)121-95(158)68(35-39-84(144)145)123-92(155)60(8)122-102(165)78(55-140)134-98(161)71(38-42-87(150)151)126-101(164)74(53-88(152)153)128-96(159)69(36-40-85(146)147)124-91(154)59(7)120-83(143)54-119-94(157)73(52-82(115)142)130-104(167)80-29-21-47-139(80)110(173)76(50-64-31-33-65(141)34-32-64)132-107(170)89(57(3)4)135-99(162)67(27-16-17-43-113)127-106(169)90(58(5)6)136-105(168)81-30-19-45-137(81)108(171)66(114)26-18-44-118-112(116)117/h10-15,22-25,31-34,56-61,66-81,89-90,140-141H,16-21,26-30,35-55,113-114H2,1-9H3,(H2,115,142)(H,119,157)(H,120,143)(H,121,158)(H,122,165)(H,123,155)(H,124,154)(H,125,163)(H,126,164)(H,127,169)(H,128,159)(H,129,166)(H,130,167)(H,131,156)(H,132,170)(H,133,160)(H,134,161)(H,135,162)(H,136,168)(H,144,145)(H,146,147)(H,148,149)(H,150,151)(H,152,153)(H,174,175)(H4,116,117,118)

InChI key

ZYDMZKPAPSZILB-UHFFFAOYSA-N

Related Categories

Amino Acid Sequence

Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

General description

The single peptide or protein standard is used to calibrate and test matrix assisted laser desorption ionization (MALDI) mass spectrometers. Adrenocorticotropic hormone (ACTH) is synthesized from its precursor pro-opiomelanocortin (POMC). It is released from anterior pituitary gland in response to corticotropin-releasing hormone (CRH) from hypothalamus.

Application

Peptide calibration standard for MALDI-MS instruments; 1 vial contains 10 nmols.
ProteoMass ACTH fragment 18-39 MALDI-MS standard has been used as a peptide calibration standard for matrix-assisted laser desorption/ionization (MALDI)-mass spectrometry (MS) instruments.

Biochem/physiol Actions

Adrenocorticotropic hormone (ACTH) stimulates the synthesis and release of glucocorticoids and androgens through cAMP-dependent by a G-protein coupled receptor in adrenal gland. Overproduction of ACTH leads to marked skin pigmentation.

Storage and Stability

Each standard is stored at room temperature and is shipped at ambient temperature. Standard stock solutions can be frozen in aliquots, but should not be subjected to more than 3 freeze-thaw cycles. Standard solutions are recommended for use for no longer than one month after reconstitution.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salivary peptidome profiling for diagnosis of severe early childhood caries.
Sun X, et al.
Journal of Translational Medicine, 14(1), 240-240 (2016)
Patient Education: A Practical Approach (2001)
Endocrinology (2001)
Attila Karsi et al.
General and comparative endocrinology, 137(3), 312-321 (2004-06-18)
Channel catfish (Ictalurus punctatus) proopiomelanocortin (POMC) cDNA was cloned to investigate its structure, evolution, and expression in different tissues. POMC is an important gene in the hypothalamus-pituitary-adrenal axis, the main mediator of the stress response. POMC gene was isolated from
Simon Ekström et al.
Journal of mass spectrometry : JMS, 42(11), 1445-1452 (2007-10-26)
A polymer microfabricated proteomic sample preparation and MALDI MS sample presentation device, the integrated selective enrichment target (ISET), comprising an array of perforated nanovials is reported. Each perforated nanovial can be filled with selective extraction media (microbeads) for purification and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service