Skip to Content
Merck
All Photos(2)

Key Documents

916870

Sigma-Aldrich

4-(1-(tert-butoxycarbonyl)piperidin-4-yl)benzoic acid

≥95%

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
MDL number:
UNSPSC Code:
12352106

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(N(CC1)CCC1C(C=C2)=CC=C2C(O)=O)OC(C)(C)C

InChI

1S/C17H23NO4/c1-17(2,3)22-16(21)18-10-8-13(9-11-18)12-4-6-14(7-5-12)15(19)20/h4-7,13H,8-11H2,1-3H3,(H,19,20)

InChI key

YCNVQGGUCDVTIZ-UHFFFAOYSA-N

Application

4-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weilin Sun et al.
Journal of medicinal chemistry, 62(6), 3122-3134 (2019-03-16)
Imatinib mesylate, 1a, inhibits production of β-amyloid (Aβ) peptides both in cells and in animal models. It reduces both the β-secretase and γ-secretase cleavages of the amyloid precursor protein (APP) and mediates a synergistic effect, when combined with a β-secretase
Andrei A Krysko et al.
Bioorganic & medicinal chemistry letters, 26(7), 1839-1843 (2016-02-26)
A series of 2-piperazin-1-yl-quinazolines were synthesized and evaluated for their antiaggregative activity. The synthesized small molecule compounds have potently inhibited platelet aggregation in vitro and blocked FITC-Fg binding to αIIbβ3 integrin in a suspension of washed human platelets. The key
Dominik K Kölmel et al.
ACS combinatorial science, 21(8), 588-597 (2019-07-10)
A new catalytic manifold that merges photoredox with nickel catalysis in aqueous solution is presented. Specifically, the combination of a highly active, yet air-stable, nickel precatalyst with a new electron-deficient pyridyl carboxamidine ligand was key to the development of a
Dillon T Flood et al.
Journal of the American Chemical Society, 141(25), 9998-10006 (2019-05-29)
DNA Encoded Libraries have proven immensely powerful tools for lead identification. The ability to screen billions of compounds at once has spurred increasing interest in DEL development and utilization. Although DEL provides access to libraries of unprecedented size and diversity
Andrei A Krysko et al.
Bioorganic & medicinal chemistry letters, 21(19), 5971-5974 (2011-08-20)
The novel RGD mimetics with phthalimidine central fragment were synthesized with the use of 4-piperidine-4-yl-butyric, 4-piperidine-4-yl-benzoic, 4-piperazine-4-yl-benzoic and 1,2,3,4-tetrahydroisoquinoline-7-carboxylic acids as surrogates of Arg motif. The synthesized compounds potently inhibited platelet aggregation in vitro and blocked FITC-Fg binding to α(IIb)β(3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service