Skip to Content
Merck
All Photos(2)

Key Documents

916846

Sigma-Aldrich

2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid

95%

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
MDL number:
UNSPSC Code:
12352106

Quality Level

Assay

95%

form

powder

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(N(CC1)CCC1C(C=CC=C2)=C2C(O)=O)OC(C)(C)C

InChI

1S/C17H23NO4/c1-17(2,3)22-16(21)18-10-8-12(9-11-18)13-6-4-5-7-14(13)15(19)20/h4-7,12H,8-11H2,1-3H3,(H,19,20)

InChI key

NPKPUCNATSURJQ-UHFFFAOYSA-N

Application

2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Base- and Additive-Free Ir-Catalyzed ortho-Iodination of Benzoic Acids: Scope and Mechanistic Investigations
Erbing E, et al.
ACS Catalysis, 8(2), 920-925 (2018)
Design and synthesis of a new class of malonyl-CoA decarboxylase inhibitors with anti-obesity and anti-diabetic activities.
Tang H, et al.
Bioorganic & Medicinal Chemistry Letters, 20(20), 6088-6092 (2010)
Visible-light-induced intramolecular sp3 C-H oxidation of 2-alkyl-substituted benzamides for the synthesis of functionalized iminoisobenzofurans..
Wu L, et al.
Chemical Communications (Cambridge, England), 55, 13908-13911 (2019)
Synthesis and Antimicrobial Evaluation of Amixicile-Based Inhibitors of the Pyruvate-Ferredoxin Oxidoreductases of Anaerobic Bacteria and Epsilonproteobacteria
Kennedy A J, et al.
Antimicrobial Agents and Chemotherapy, 60(7), 3980-3987 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service