Skip to Content
Merck
All Photos(1)

Key Documents

91312

Sigma-Aldrich

(R)-(−)-Phenylephrine hydrochloride

tested according to Ph. Eur.

Synonym(s):

Phenylephrini hydrochloridum, (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
4158948
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

form

powder

mp

143-145 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

OC1=CC([C@H](CNC)O)=CC=C1.Cl

InChI

1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m0./s1

InChI key

OCYSGIYOVXAGKQ-FVGYRXGTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale A Currigan et al.
Anesthesiology, 121(5), 930-936 (2014-09-10)
Vasopressor drugs, commonly used to treat systemic hypotension and maintain organ perfusion, may also induce regional vasoconstriction in specialized vascular beds such as the lung. An increase in pulmonary vascular tone may adversely affect patients with pulmonary hypertension or right
C A Boly et al.
British journal of anaesthesia, 111(5), 750-758 (2013-07-11)
Left-ventricular end-systolic elastance (Ees) is an index of cardiac contractility, but the invasive nature of its assessment has limited perioperative application. We explored the feasibility of a minimally invasive method of Ees estimation for perioperative assessment of cardiac function and
Stephanie W Watts et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(6), 1320-1328 (2013-04-06)
Obesity and hypertension are comorbid in epidemic proportion, yet their biological connection is largely a mystery. The peptide chemerin is a candidate for connecting fat deposits around the blood vessel (perivascular adipose tissue) to arterial contraction. We presently tested the
Edmund Tsui et al.
Experimental eye research, 112, 134-138 (2013-05-25)
The impact of mydriatic agents on the standardized provocation of retinal vascular reactivity has not been systematically investigated. Our aim was to investigate the effect of commonly used mydriatic agents on the provoked vascular response of retinal arterioles. One eye
Thomas R Sundermeier et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(8), 3780-3791 (2014-05-09)
Photoreceptor cell death is the proximal cause of blindness in many retinal degenerative disorders; hence, understanding the gene regulatory networks that promote photoreceptor survival is at the forefront of efforts to combat blindness. Down-regulation of the microRNA (miRNA)-processing enzyme DICER1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service