Skip to Content
Merck
All Photos(1)

Key Documents

91036

Supelco

β-Ionone

analytical standard

Synonym(s):

4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C(C)\C=C\C1=C(CCCC1(C)C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

General description

β-Ionone, an end ring analog of β-carotene,[1] is a volatile carotenoid that contributes to the fragrance in many flowers, including petunia (Petunia hybrida).[2]

Application

β-Ionone may be used as a reference standard in the determination of β-ionone in wine using solid-phase extraction followed by gas chromatography with mass spectrometry (GC-MS).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circadian regulation of the PhCCD1 carotenoid cleavage dioxygenase controls emission of beta-ionone, a fragrance volatile of petunia flowers.
Simkin JA, et al.
Plant Physiology, 136(3), 3504-3514 (2004)
Selection of astaxanthin-overproducing mutants of Phaffia rhodozyma with beta-ionone.
Lewis MJ, et al.
Applied and Environmental Microbiology, 56(9), 2944-2945 (1990)
Determination of minor and trace volatile compounds in wine by solid-phase extraction and gas chromatography with mass spectrometric detection.
Lopez, et al.
Journal of Chromatography A, 966(1-2), 167-177 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service