Skip to Content
Merck
All Photos(1)

Documents

16976

Supelco

β-Cyclocitral

analytical standard

Synonym(s):

2,6,6-Trimethyl-1-cyclohexene-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.497 (lit.)
n20/D 1.497-1.501

bp

62-63 °C/3 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C1=C(C)CCCC1(C)C

InChI

1S/C10H16O/c1-8-5-4-6-10(2,3)9(8)7-11/h7H,4-6H2,1-3H3

InChI key

MOQGCGNUWBPGTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Cyclocitral is a volatile compound, a C7 oxidized derivative of β-carotene. It is an analog of safranal that contributes to the flavor/aroma of several fruits, vegetables, ornamental plants, and also helps in attracting pollinators.

Application

β-Cyclocitral may be used as an analytical standard for the determination of the analyte in Sicilian (South Italy) saffron, natural water body and selected strains of Cyanobacteria by chromatography-based techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alexandra J Dickinson et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(21), 10563-10567 (2019-05-10)
Natural compounds capable of increasing root depth and branching are desirable tools for enhancing stress tolerance in crops. We devised a sensitized screen to identify natural metabolites capable of regulating root traits in Arabidopsis β-Cyclocitral, an endogenous root compound, was
Determination of volatile organic compounds in selected strains of cyanobacteria
Milovanovic I, et al.
Journal of Chemistry, 2015(24) (2015)
Bioactive volatiles in Sicilian (South Italy) saffron: Safranal and its related compounds
Condurso C, et al.
J. Essent. Oil Res., 29(3), 221-227 (2017)
Dihydroactinidiolide, a high light-induced $\beta$-carotene derivative that can regulate gene expression and photoacclimation in Arabidopsis
Shumbe L, et al.
Molecular plant, 7(7), 1248-1251 (2014)
Xuwei Deng et al.
Journal of chromatography. A, 1218(24), 3791-3798 (2011-05-14)
Production and fate of taste and odor (T&O) compounds in natural waters are a pressing environmental issue. Simultaneous determination of these complex compounds (covering a wide range of boiling points) has been difficult. A simple and sensitive method for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service