Skip to Content
Merck
All Photos(1)

Key Documents

W259500

Sigma-Aldrich

β-Ionone

predominantly trans, ≥97%, FCC, FG

Synonym(s):

4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
2595
Beilstein:
1909544
Council of Europe no.:
142
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.008
NACRES:
NA.21
Organoleptic:
berry; woody; floral; sweet
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.52 (lit.)

bp

126-128 °C/12 mmHg (lit.)

density

0.945 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; woody; floral; sweet

SMILES string

CC(=O)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Ionone is one of the main volatile compounds identified in green tea and tomatoes.

Biochem/physiol Actions

Taste at 1-20 ppm

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic modification of tomato homogenate and its effect on volatile flavor compounds.
Yilmaz E, et al.
Journal of Food Science, 67(6), 2122-2125 (2002)
Further Investigation of Flavor Constituents in Manufactured Green Tea
Yamanishi T, et al.
Agricultural and Biological Chemistry, 34(4), 599-608 (1970)
Jianfeng Wang et al.
Investigative ophthalmology & visual science, 53(10), 6355-6369 (2012-08-18)
In human retinal degeneration, rod photoreceptors reactively sprout neurites. The mechanism is unknown in part because of the paucity of animal models displaying this feature of human pathology. We tested the role of cAMP and opsin in sprouting by tiger
A Vieira et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 44(6), 538-545 (2011-03-30)
β-ionone (βI), a cyclic isoprenoid, and geraniol (GO), an acyclic monoterpene, represent a promising class of dietary chemopreventive agents against cancer, whose combination could result in synergistic anticarcinogenic effects. The chemopreventive activities of βI and GO were evaluated individually or
Sivakumar Sekharan et al.
Journal of the American Chemical Society, 132(45), 15856-15859 (2010-10-23)
Visual pigment rhodopsin provides a decisive crossing point for interaction between organisms and environment. Naturally occurring visual pigments contain only PSB11 and 3,4-dehydro-PSB11 as chromophores. Therefore, the ability of visual opsin to discriminate between the retinal geometries is investigated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service