Skip to Content
Merck
All Photos(1)

Documents

49085

Supelco

Atrazine

analytical standard

Synonym(s):

2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
Beilstein:
612020
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

CCNc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)

InChI key

MXWJVTOOROXGIU-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Atrazine is a commonly used herbicide, which belongs to the triazine class of herbicides. It is generally used for weed control management in agricultural crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Hermaphroditic, demasculinized frogs after exposure to the herbicide atrazine at low ecologically relevant doses
Hayes.BT, et al.
Proceedings of the National Academy of Sciences of the USA, 99, 5476-5480 (2002)
Ecological risk assessment of atrazine in North American surface waters
Solomon.RK, et al.
Environmental Toxicology and Chemistry / Setac, 15, 31-76 (1996)
Karsten Nödler et al.
Marine pollution bulletin, 70(1-2), 112-118 (2013-03-14)
Despite its EU-wide ban in 2004 the triazine herbicide atrazine (ATR) and its transformation products (TPs) are frequently detected in the aqueous environment. In this study, concentrations of ATR, two of its TPs, and terbuthylazine (TERB) are presented and discussed
Keith R Solomon et al.
Critical reviews in toxicology, 38(9), 721-772 (2008-10-23)
The herbicide atrazine is widely used in agriculture for the production of corn and other crops. Because of its physical and chemical properties, atrazine is found in small concentrations in surface waters--habitats for some species. A number of reports on
M Graymore et al.
Environment international, 26(7-8), 483-495 (2001-08-04)
A portion of all herbicides applied to forests, croplands, road sides, and gardens are inevitably lost to water bodies either directly through runoff or indirectly by leaching through groundwater into ephemeral streams and lakes. Once in the aquatic environment, herbicides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service