Skip to Content
Merck
All Photos(1)

Key Documents

89720

Sigma-Aldrich

Sodium p-toluenesulfinate

purum, anhydrous, ≥96.0% (NT)

Synonym(s):

p-Toluenesulfinic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-CH3-C6H4SO2Na
CAS Number:
Molecular Weight:
178.18
Beilstein:
3574491
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

purum

Assay

≥96.0% (NT)

form

powder

functional group

sulfinic acid

SMILES string

[Na+].Cc1ccc(cc1)S([O-])=O

InChI

1S/C7H8O2S.Na/c1-6-2-4-7(5-3-6)10(8)9;/h2-5H,1H3,(H,8,9);/q;+1/p-1

InChI key

KFZUDNZQQCWGKF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paratchata Batsomboon et al.
The Journal of organic chemistry, 74(10), 4009-4012 (2009-04-24)
2-Arylchromans were readily prepared from the hetero-Diels-Alder reactions of styrenes with the ortho-quinone methides (o-QMs) which, in turn, were generated by treating the MOM-protected benzylacetate derivatives with p-TsOH immobilized on silica (PTS-Si) in toluene under mild conditions (0 degrees C
Véronique Fournier et al.
Journal of chromatography. A, 1129(1), 21-28 (2006-08-09)
Addition of long-chain polyunsaturated fatty acids (LC-PUFAs) from marine oil into food products implies preliminary refining procedures of the oil which thermal process affects the integrity of LC-PUFAs. Deodorization, the major step involving high temperatures, is a common process used
Pierluigi Delmonte et al.
Journal of chromatography. A, 1214(1-2), 30-36 (2008-11-14)
In recent years, several countries have implemented new regulations regarding limitations or labeling of the trans fatty acid (tFA) content in foods. In order to comply with the new requirements, gas chromatographic methods for fatty acid (FA) analysis have been
Synthesis of p-toluenesulfinic esters of cellulose and beta-cyclodextrin.
C Roussel et al.
Carbohydrate research, 282(2), 307-313 (1996-03-18)
Shirin Safaei et al.
Molecular diversity, 16(3), 591-600 (2012-08-29)
A variety of pyrazolo[3,4-d]pyrimidine-6(7H)-thione derivatives were easily synthesized with a novel, simple, efficient, and regioselective method via three-component condensation reaction of 5-methyl-1H-pyrazol-3-amine, arylisothiocyanates, and aldehydes in the presence of catalytic amount of p-toluenesulfonic acid (p-TSA) in 1-butyl-3-methylimidazolium bromide ionic liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service