88431
Ammonium phosphate monobasic
analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Synonym(s):
mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate
About This Item
Recommended Products
grade
analytical standard
Quality Level
description
traceable to NIST SRM 194
Assay
≥99.5% (T)
≥99.5%
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
titration: suitable
impurities
12.18%±0.10% total Kjeldahl nitrogen
loss
≤0.2% loss on drying
pH
3.8-4.4 (25 °C, 50 mg/mL in H2O)
mp
190 °C (dec.) (lit.)
solubility
H2O: soluble, clear, colorless
application(s)
food and beverages
format
neat
SMILES string
N.OP(O)(O)=O
InChI
1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)
InChI key
LFVGISIMTYGQHF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Available in a neat format, in a secure glass bottle to ensure its stability for the entire shelf life until opened.
- Offers accurate titer determinations
- Accompanied by a certificate of analysis (CoA)
Caution
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service