Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium phosphate monobasic
Synonym(s):
mono
-Ammonium phosphate,
prim
-Ammonium phosphate, Ammonium dihydrogenphosphate
Linear Formula:
NH
4
H
2
PO
4
CAS Number:
7722-76-1
Molecular Weight:
115.03
EC Number:
231-764-5
Products
Product Category
salts (5)
bases (1)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
216003
Ammonium phosphate monobasic,
ACS reagent, ≥98%
Expand
View Pricing
A1645
Ammonium phosphate monobasic,
ReagentPlus
®
, ≥98.5%
Expand
View Pricing
467782
Ammonium phosphate monobasic,
≥99.99% trace metals basis
Expand
View Pricing
A3048
Ammonium phosphate monobasic,
suitable for plant cell culture
Expand
View Pricing
09709
Ammonium phosphate monobasic,
BioUltra, ≥99.5% (T)
Expand
View Pricing
795461
Ammonium phosphate monobasic,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥98%
Expand
View Pricing
88431
Ammonium phosphate monobasic,
analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Expand
View Pricing