Skip to Content
Merck
All Photos(1)

Key Documents

86476

Supelco

γ-Terpinene

analytical standard

Synonym(s):

1-Isopropyl-4-methyl-1,4-cyclohexadiene, p-Mentha-1,4-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2038347
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.7 (vs air)

vapor pressure

~0.7 mmHg ( 20 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.474 (lit.)
n20/D 1.474

bp

182 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)C1=CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,7-8H,5-6H2,1-3H3

InChI key

YKFLAYDHMOASIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Terpinene is a monoterpene hydrocarbon present in essential oils extracted from Thymus vulgaris L., and exhibits antimicrobial, carminative and expectorant activities.

Application

γ-Terpinene has been used as a standard in the determination of γ-terpinene, present in tea tree oil extracted from various parts of Melaleuca alternifolia tree and analyzed using gas chromatography coupled with mass spectrometry(GC-MS).
γ-Terpinene may be used as an analytical reference standard for the quantification of the analyte in Thymus vulgaris L. oil samples using gas chromatography coupled to mass spectrometry (GC-MS).
γ–terpinene has been used as a standard in the determination of γ–terpinene, present in tea tree oil extracted from various parts of Melaleuca alternifolia tree and analyzed using gas chromatography coupled with mass spectrometry(GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: thymus

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

124.9 °F - closed cup

Flash Point(C)

51.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC/MS evaluation of thyme (Thymus vulgaris L.) oil composition and variations during the vegetative cycle
Hudaib M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(4), 691-700 (2002)
Correlations of the components of tea tree oil with its antibacterial effects and skin irritation
Lee J-C, et al.
Journal of food and drug analysis, 21, 169-176 (2013)
Nataraj Jagannath et al.
Natural product communications, 7(7), 943-946 (2012-08-23)
The essential oil was extracted from the seeds of Heracleum rigens by hydrodistillation and a total of twenty compounds accounting for 98.5% of the total oil composition were identified. Physicochemical properties and chemical composition of the oil was determined by
Ram Swaroop Verma et al.
Natural product research, 26(14), 1358-1362 (2011-10-21)
Essential oils derived from six different phenophases, namely early vegetative stage, late vegetative stage, early flowering stage, full flowering stage (FFS), late flowering stage and seed shattering stage of Origanum vulgare L. grown in Kumaon region of Uttarakhand, India were
R S Verma et al.
Natural product research, 24(20), 1890-1896 (2010-11-26)
Thymus linearis (Benth. ex Benth) was collected from five distinct locations of western Himalaya (India) during the summer season. The hydro-distilled essential oil (yield 0.84-0.95%) was analysed by gas chromatography (GC) and gas chromatography-mass spectrometry (GC-MS). A total of 56

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service