Skip to Content
Merck
All Photos(1)

Key Documents

64643

Supelco

Myrcene

analytical standard

Synonym(s):

β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
136.23
Beilstein:
1719990
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.7 (vs air)

vapor pressure

~7 mmHg ( 20 °C)

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.469 (lit.)

bp

167 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C/CCC(=C)C=C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7H,1,4,6,8H2,2-3H3

InChI key

UAHWPYUMFXYFJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myrcene is a naturally occurring volatile monoterpene found in various plants, such as wild thyme, lemon grass, mango, cardamom, and cannabis. It is widely used as an aroma chemical in personal care and household products, besides being used as a flavoring agent (food additive) in various foods and beverages.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Application

This analytical standard can also be used as follows:
  • Determination of four flavor compounds— isoamyl acetate, ethyl hexanoate, benzaldehyde, and myrcene, in commercial beer samples by using two solvent-less sample treatment techniques of stir-bar sorptive extraction (SBSE) and solid-phase microextraction (SPME) for their subsequent analysis by gas chromatography-flame ionization detection (GC-FID)
  • Analysis of fresh mano samples for the detection and quantification of myrcene using a quartz crystal microbalance (QCM) sensor, modified with ethyl cellulose
  • Multi-residue analysis of volatiles and fatty acids found in wild and cultivated fennel samples by a single extraction method and gas chromatographic-flame ionization detection (GC-FID)
  • Identification and determination of volatile aroma compounds, commonly present in three plant species from the Citrus genus by using simultaneous distillation extraction (SDE) technique for sample treatment and analysis by gas chromatography-mass spectrometry (GC-MS) in electron ionization mode (EI)
  • Secondary metabolite profiling of various plant parts collected from 82 plants belonging to 21 different cannabis strains using gas chromatography-mass spectrometry (GC-MS) for sterols and terpenoids (mono-, sesqui-, tri-), and high-performance liquid chromatography (HPLC) with UV and mass spectrometric (MS) detection for flavonoids

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: thymus

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro inhibition of CYP2B1 monooxygenase by ?-myrcene and other monoterpenoid compounds
De-Oliveira X.A.CA, et al.
Toxicology Letters, 92(1), 39-46 (1997)
Flavia Bonamin et al.
Chemico-biological interactions, 212, 11-19 (2014-02-01)
The monoterpene β-myrcene has been widely used in cosmetics, food and beverages, and it is normally found in essential oil from citrus fruit. The aim of this study was to investigate the anti-ulcer effects of β-myrcene on experimental models of
Mark L Thompson et al.
Applied microbiology and biotechnology, 85(3), 721-730 (2009-08-27)
The biocatalytic generation of high-value chemicals from abundant, cheap and renewable feedstocks is an area of great contemporary interest. A strain of Rhodococcus erythropolis designated MLT1 was isolated by selective enrichment from the soil surrounding hop plants, using the abundant
A Rivoal et al.
Chemosphere, 78(8), 942-949 (2010-01-23)
The large amount of volatile organic compound (VOC) emitted by vegetation modifies air quality contributing to both tropospheric ozone and secondary organic aerosol production. A better understanding of the factors controlling VOC emissions by vegetation is mandatory in order to
Johannes Stökl et al.
Proceedings. Biological sciences, 278(1709), 1216-1222 (2010-10-15)
Most insects are dependent on chemical communication for activities such as mate finding or host location. Several plants, and especially orchids, mimic insect semiochemicals to attract insects for unrewarded pollination. Here, we present a new case of pheromone mimicry found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service