Skip to Content
Merck
All Photos(3)

Documents

114359

Sigma-Aldrich

Bromocresol Green

ACS reagent, Dye content 95 %

Synonym(s):

BCG; 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1λ6-benzoxathiol-3-yl]-3-methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Br4O5S
CAS Number:
Molecular Weight:
698.01
Beilstein:
372527
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

Assay

≥95% (After Oxygen Combustion, silver nitrate titration)

form

powder

composition

Dye content, 95%

visual transition interval

3.8, yellow(passes test)
3.8-5.4, yellow to blue(passes test)
4.5, green
5.4, blue

mp

225 °C (dec.) (lit.)

λmax

423 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1c(Br)c(O)c(Br)cc1C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4C

InChI

1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3

InChI key

FRPHFZCDPYBUAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromocresol Green is a synthetic triphenylmethane dye that is anionic in nature. At pH 3.8-5.4, it changes color from yellow to blue-green, and is therefore, commonly used as a pH indicator.

Application

Bromocresol green serves as a pH indicator in biology and medicinal experiments.
  • It is used widely as a vital stain to study blood-brain permeability.
  • It has been used in the pH-sensitive colorimetric determination of glutamate decarboxylase activity.
  • It has also been employed in the evaluation of paper-analytical devices for colorimetric analysis of serum samples.
  • It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations, and in charge-transfer complexation processes.
  • It is used in TLC for the visualization of compounds with functional groups whose pKa is below 5.0.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Chitosan Poly(methacrylate) Composites for Adsorption of Bromocresol Green
Liu, et al.
ACS Omega, 4 (2019)
A high-throughput colorimetric assay to measure the activity of glutamate decarboxylase
Yu, et al.
Enzyme and Microbial Technology, 49 (2011)
Martina O' Toole et al.
Talanta, 66(5), 1340-1344 (2008-10-31)
A novel inexpensive optical-sensing technique has been developed for colorimetric flow analysis. This sensing system employs two LEDs whereby one is used as the light source and the other as a light detector. The LED used as light detector is
Conn?s Biological Stains: A Handbook of Dyes, Stains and Fluorochromes for Use in Biology and Medicine (10th ed.)
Horobin, R. W., & Kiernan, J. A. (Eds.)
Conn?s Biological Stains (2002)
Selection of appropriate protein assay method for a paper microfluidics platform
Pokhrel, et al.
practical laboratory medicine, 21 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service