Skip to Content
Merck
All Photos(5)

Key Documents

N7005

Sigma-Aldrich

Neutral Red

Dye content ≥90 %, Powder

Synonym(s):

3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Basic Red 5, Toluylene red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
Colour Index Number:
50040
Beilstein:
3918740
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Neutral Red, Dye content ≥90 %

form

powder

Quality Level

composition

Dye content, ≥90%

technique(s)

titration: suitable

color

brown to black

pH

6.8-8.0, red to yellow

mp

290 °C (dec.) (lit.)

solubility

water: 10 mg/mL, dark red

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Cl.CN(C)c1ccc2nc3cc(C)c(N)cc3nc2c1

InChI

1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H

InChI key

PGSADBUBUOPOJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neutral red (C.I. 50040) is a protonated diaminophenazine dye. It is aminodimethylaminomethylphenazinehydrochloride, C15H17N4Cl.

Application

Neutral red has multiple staining applications that include its use as a nuclear stain, counterstain in the Gram procedure, either alone, or as a component of the Twort stain (a fast green-neutral red mixture). Neutral red staining is also a viability assessment method used for the detection of viable yeast cells in dandruff samples and of mussel larvae. Neutral red is also recommended for use in viability assessment as it is selectively absorbed by lysosomes and other acidic organelles in cells. It is also used for supravital staining of living blood cells.
Useful as an indicator for preparing neutral red paper, and as a biological stain.

Biochem/physiol Actions

Neutral red is a widely used cationic fluorochrome that is known to accumulate in the Golgi complex and nuclei after first staining lysosomes.

Quality

Purified

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chlorophyll derivative mediated PDT versus methotrexate: an in vitro study using MCF-7 cells.
Gomaa I
Photodiagnosis and Photodynamic Therapy, 9, 362-362 (2012)
Effects of hypocretin2-saporin and antidopamine-beta-hydroxylase-saporin neurotoxic lesions of the dorsolateral pons on sleep and muscle tone.
Blanco-Centurion C
The European Journal of Neuroscience, 19, 2741-2741 (2004)
Sabnis RW
Handbook of Acid-Base Indicators (2007)
Cell Growth, Differentiation and Senescence: A Practical Approach (1999)
Mara Ceridono et al.
Regulatory toxicology and pharmacology : RTP, 63(3), 480-488 (2012-06-13)
This is the report from the "ECVAM-EFPIA workshop on 3T3 NRU Phototoxicity Test: Practical Experience and Implications for Phototoxicity Testing", jointly organized by ECVAM and EFPIA and held on the 25-27 October 2010 in Somma Lombardo, Italy. The European Centre

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service