Skip to Content
Merck
All Photos(1)

Key Documents

81512

Supelco

Poly(methyl methacrylate)

analytical standard, for GPC, 8,000

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~7910
Mp ~8350
Mw ~8290

technique(s)

gel permeation chromatography (GPC): suitable

solubility

H2O: insoluble

density

1.200 g/cm3

Mw/Mn

~1.05

format

neat

storage temp.

2-8°C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucía D Garza-García et al.
Lab on a chip, 13(7), 1243-1246 (2013-02-16)
We report a proof-of-principle for the use of micro-devices as continuous bioreactors for the production of monoclonal antibodies. We culture CHO cells on the surface of PMMA "zigzag" channels textured with semi-spherical cavities coated with fibronectin, observing steady-state productivities 100
Pleural infection 55 years after plombage for pulmonary tuberculosis.
Qian-kun Chen et al.
Asian cardiovascular & thoracic annals, 21(1), 100-100 (2013-02-23)
S A Bini et al.
The bone & joint journal, 95-B(3), 367-370 (2013-03-02)
We evaluated the impact of pre-coating the tibial component with polymethylmethacrylate (PMMA) on implant survival in a cohort of 16 548 primary NexGen total knee replacements (TKRs) in 14 113 patients. In 13 835 TKRs a pre-coated tray was used
Kariem Mostafa Al Batouty et al.
The journal of adhesive dentistry, 15(2), 161-166 (2013-03-28)
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according
Right ventricular perforation and pulmonary embolism with polymethylmethacrylate cement after percutaneous kyphoplasty.
Igor Gosev et al.
Circulation, 127(11), 1251-1253 (2013-03-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service