Skip to Content
Merck
All Photos(1)

Key Documents

81501

Supelco

Poly(methyl methacrylate)

analytical standard, for GPC, 50,000

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~54000
Mp ~56900
Mw ~55900

technique(s)

gel permeation chromatography (GPC): suitable

solubility

H2O: insoluble

density

1.200 g/cm3

Mw/Mn

~1.03

format

neat

storage temp.

2-8°C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(methyl methacrylate) is a material which can be used in the fabrication of micro-fluid chips.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and spectral characterization of poly (methyl methacrylate)/hydroxyapatite composite for biomedical applications
Chen Y, et al.
Development and spectral characterization of poly (methyl methacrylate)/hydroxyapatite composite for biomedical applications, 18(1), 41-45 (2004)
Gursukhman S Sidhu et al.
Journal of neurosurgery. Spine, 19(1), 61-70 (2013-05-07)
The authors endeavor to highlight the surgical management of severe neurological deficit resulting from cement leakage after percutaneous vertebroplasty and to systematically review the literature on the management of this complication. A patient presented after a vertebroplasty procedure for traumatic
Pleural infection 55 years after plombage for pulmonary tuberculosis.
Qian-kun Chen et al.
Asian cardiovascular & thoracic annals, 21(1), 100-100 (2013-02-23)
S A Bini et al.
The bone & joint journal, 95-B(3), 367-370 (2013-03-02)
We evaluated the impact of pre-coating the tibial component with polymethylmethacrylate (PMMA) on implant survival in a cohort of 16 548 primary NexGen total knee replacements (TKRs) in 14 113 patients. In 13 835 TKRs a pre-coated tray was used
Kariem Mostafa Al Batouty et al.
The journal of adhesive dentistry, 15(2), 161-166 (2013-03-28)
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service