Skip to Content
Merck
All Photos(1)

Documents

77608

Sigma-Aldrich

Phenol

BioUltra, for molecular biology, ≥99.5% (GC)

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

solid

autoignition temp.

1319 °F

contains

~0.15% hypophosphorous acid as stabilizer

expl. lim.

8.6 %

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.5% water

ign. residue

≤0.05%

pH

3.0-6.0 (25 °C, 0.5 M in H2O)

bp

182 °C (lit.)

mp

40-42 °C (lit.)
40-42 °C

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

1.071 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Oc1ccccc1

absorption

cut-off at 294 nm in H2O at 0.5 M

suitability

suitable for molecular biology

storage temp.

2-8°C

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenol/carbolic acid is mainly produced by hospital pharmacy as it is unavailable in premixed liquid form. It is not highly soluble in water. Phenol should be refrigerated and protected from light exposure to maintain its shelf life of approximately one year. Kidneys can excrete phenol as several conjugated derivatives.

Application

Phenol has been used in the incubation of pellet for RNA extraction.

Biochem/physiol Actions

Phenol has the ability to denature protein, hence can lead to denervation. At lower concentration, it can serve as a local anaesthetic and can also act as a neurolytic agent in higher concentration. It is also linked with tissue damage at higher concentrations.

Other Notes

For the preparation of DNA ; Large- and small-scale phenol extractions

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical neurolytic blocks
Practical Management of Pain, 784-793 (2013)
Commonly Used Medications in Procedures
Pain Procedures in Clinical Practice, 2-2 (2011)
Neural blockade with neurolytic agents
Pain management, 343-348 (2007)
Current Protocols in Molecular Biology (1987)
Telomere length determines TERRA and R-loop regulation through the cell cycle
Graf M, et al.
Cell, 170(1), 72-85 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service