Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
8.22296
Phenol
for synthesis
Synonym(s):
Hydroxybenzene, Carbolic acid, Hydroxybenzene
About This Item
0.36 mmHg ( 20 °C)
grade
for synthesis
Quality Level
vapor density
3.24 (vs air)
vapor pressure
0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)
Assay
≥99% (GC)
form
crystalline solid
autoignition temp.
1319 °F
potency
660 mg/kg LD50, skin (Rat)
expl. lim.
8.6 %
dilution
(for synthesis)
pH
5 (20 °C, 50 g/L in H2O)
bp
182 °C (lit.)
mp
40-42 °C (lit.)
transition temp
flash point 81 °C
solubility
84 g/L
density
1.071 g/mL at 25 °C (lit.)
bulk density
620 kg/m3
storage temp.
2-30°C
SMILES string
Oc1ccccc1
InChI
1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H
InChI key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Catalytic asymmetric dearomatization (CADA) reactions of phenol and aniline derivatives: This review highlights the use of phenols in catalytic asymmetric dearomatization, presenting the latest advancements in this field. (WT Wu, L Zhang, SL You - Chemical Society Reviews, 2016 - rsc.org).
- External oxidant-free electrooxidative annulation between phenol and indole derivatives: Explores the electrooxidative coupling of phenols and indoles in the presence of various substituents, highlighting the method′s efficiency without external oxidants. (K Liu, S Tang, P Huang, A Lei - Nature Communications, 2017 - nature.com).
Analysis Note
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 43 °C
Identity (IR): passes test
Protect from light. Any pink discoloration that might result from oxidation during storage generally does not affect the quality of the product.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2
Target Organs
Nervous system,Kidney,Liver,Skin
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service