73426
Nitrate Reduction Test
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171621
NACRES:
NA.85
Recommended Products
product line
BioChemika
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
microbe id | metabolite detection: suitable
application(s)
agriculture
clinical testing
environmental
food and beverages
pharmaceutical
microbiology
storage temp.
2-8°C
suitability
Enterobacter spp.
Neisseria spp.
anaerobic bacteria
bacteria
General description
Bacterial species may be differentiated on the basis of their ability to reduce nitrate to nitrite or nitrogenous gases. The reduction of nitrate may be coupled to anaerobic respiration in some species.
Components
Nitrate broth (Cat. No. 72548) 100 g
Sulfanilic acid (Cat. No. 86090) = Reagent A 100 g,
N,N-Dimethyl-1-naphthylamine (Sial D4011) = Reagent B 10 mL
Zinc (Cat. No. 93027) 1 g.
Sulfanilic acid (Cat. No. 86090) = Reagent A 100 g,
N,N-Dimethyl-1-naphthylamine (Sial D4011) = Reagent B 10 mL
Zinc (Cat. No. 93027) 1 g.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Ox. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
5.1B - Oxidizing hazardous materials
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amelia Tang et al.
Microorganisms, 8(3) (2020-04-05)
In the midst of the major soil degradation and erosion faced by tropical ecosystems, rehabilitated forests are being established to avoid the further deterioration of forest lands. In this context, cellulolytic, nitrogen-fixing (N-fixing), phosphate-solubilizing bacteria are very important functional groups
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service