71037BC
Assurance® GDS EHEC ID for E. coli O157:H7 Kit
suitable for PCR testing in Food and Beverages applications
About This Item
Recommended Products
Product Name
GDS EHEC ID for E.coli O157:H7 TQ,
Agency
PCR, Immunomagnetic Separation (IMS)
according to AOAC® (PTM)
Quality Level
shelf life
6‑12 mo.
manufacturer/tradename
BioControl Systems
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
BioControl GDS
for use with GDS Rotor-Gene
storage temp.
2-8°C
suitability
Escherichia coli (O157:H7)
Related Categories
General description
Application
Features and Benefits
- Designed specifically to follow the GDS E. coli O157:H7 Tq assay
- Results available as fast as 12 hours for composite 375 g samples
- Single harmonized proprietary mEHEC enrichment
- Validated for raw ground beef, raw beef trim, finely textured beef (FTB), and carcass cloths
- Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and fast time to results
Components
- EHEC ID Amplification Tubes
- O157 Concentration Reagent
- Resuspension Buffer Tq
- Wash solution
Testing Method
Legal Information
also commonly purchased with this product
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service