Skip to Content
Merck

Skip To

73071BC

GDS Rotor-Gene Q Laptop

Laptop for use with Assurance® GDS Rotor-Gene® Q thermocycler

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41103909
NACRES:
NC.81
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

Assurance GDS®, BioControl Systems 73071

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

GDS Rotor-Gene® Q thermocycler is supported with Dell Latitude laptop. Dell Latitude laptops work faster and provides flexible, simplified and secure solutions. The laptop is made with premium finishes like carbon fiber and all new aluminum, ensuring reimagined design and durability in a lighter frame. The laptop is pre-installed with the comprehensive Rotor-Gene® Q software package that supports the state-of-the art real-time analysis procedures from basic to advanced algorithms. The software determines sample results for the user and increases the reliability of results. Data security is assured, and all process steps are trackable from starting the run to exporting the results.

Application

GDS Rotor-Gene® Q Laptop pre-installed with the software determines sample results for the user and increases the reliability of results with assured data security.
PCR

Features and Benefits

  • BioControl GDS software pre-installed
  • Optimally configured for Assurance® GDS
  • 3 User profiles:
1. The “GDS Administrator” user level provides administrators with the ability to set up new users, Password protected, updated software, and configure the computer′s operational settings.
2. The “GDS Analyst” user level provides laboratory managers with the ability to analyze data in greater detail. Analysts have the ability to generate reports that can be saved, printed, or e-mailed directly from the GDS Rotor-Gene® software and is password protected.
3. The “GDS Operator” is the default access level that allows the user to enter sample information and provides a convenient results table.

Other Notes

  • Dell Latitude laptop
  • A/C Adapter

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
71022BC79145BC71010ATMBC
compatibility

for use with GDS Rotor-Gene

compatibility

for use with GDS Rotor-Gene

compatibility

BioControl GDS

compatibility

BioControl GDS, for use with GDS Rotor-Gene

storage temp.

room temp

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

manufacturer/tradename

Assurance GDS®, BioControl Systems 73071

manufacturer/tradename

Assurance GDS®, BioControl Systems 71022-100, BioControl Systems 71022-576ATM

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79145

manufacturer/tradename

BioControl Systems 710010-576ATM

Quality Level

EQ1

Quality Level

200

Quality Level

EQ1

Quality Level

200

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters