Skip to Content
Merck
All Photos(2)

Documents

68654

Supelco

Vanillic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Hydroxy-3-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein:
2208364
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

208-210 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(ccc1O)C(O)=O

InChI

1S/C8H8O4/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,9H,1H3,(H,10,11)

InChI key

WKOLLVMJNQIZCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: angelica carum cinnamomum curcuma ginkgo humulus lycium melissa panax pimpinella thymus zingiber

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Lee et al.
Clinical chemistry, 39(9), 1788-1792 (1993-09-01)
We describe a sensitive and specific high-performance liquid-chromatographic method for determining the benzene metabolite, trans,trans-muconic acid (ttMA) in urine by measuring ultraviolet absorbance at 265 nm. We mix 1 mL of urine sample with 2 mL of Tris buffer containing
Rafael Llorach et al.
Journal of proteome research, 8(11), 5060-5068 (2009-09-17)
Cocoa-phytochemicals have been related to the health-benefits of cocoa consumption. Metabolomics has been proposed as a powerful tool to characterize both the intake and the effects on the metabolism of dietary components. Human urine metabolome modifications after single cocoa intake
Changgeng Liu et al.
Environmental science & technology, 46(24), 13262-13269 (2012-11-23)
Methoxyphenols, tracers for wood smoke, are emitted into the atmosphere in large quantities, but their chemical degradation in the atmosphere has not been well characterized. In this study, heterogeneous kinetics of particulate syringaldehyde (SA), vanillic acid (VA), and coniferyl aldehyde
F A Muskiet et al.
Clinical chemistry, 25(7), 1281-1284 (1979-07-01)
We report quantitative data on beta-glucuronidase- and sulfatase-hydrolyzable conjugates of homovanillic acid, 3,4-dihydroxyphenylacetic acid, p-hydroxyphenylacetic acid, and vanillic acid in the urine of 20 apparently normal and healthy control persons and of three patients with neuroblastoma. We used organic solvent
Zhuliang Tan et al.
Food chemistry, 138(2-3), 1438-1447 (2013-02-16)
Phytosterols and their derivatives have attracted much attention because of their health benefits to humans and are widely used in food, pharmaceuticals, and cosmetics in the past decades. While most of the research has focused on free phytosterols and phytosteryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service