Skip to Content
Merck
All Photos(4)

Documents

H20059

Sigma-Aldrich

4-Hydroxybenzoic acid

ReagentPlus®, 99%

Synonym(s):

p-HBA, p-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
970950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

mp

213-217 °C (lit.)

solubility

alcohol: freely soluble

SMILES string

OC(=O)c1ccc(O)cc1

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

FJKROLUGYXJWQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxybenzoic acid (PHBA) is a phenolic derivative of benzoic acid, used as a building block in polymer synthesis.

Application

4-Hydroxybenzoic acid can be used as a starting material to synthesize organic intermediates such as 4-acetoxybenzoic acid and 4-hydroxy-3,5-diiodobenzoic acid.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formation of novel polymeric films derived from 4-hydroxybenzoic acid
Ferreira L, et al.
Materials Chemistry and Physics, 129, 46-52 (2011)
Esterification of Carboxylic Acids with Trialkyloxonium Salts: Ethyl and Methyl 4?Acetoxybenzoates: Benzoic acid, 4?(acetyloxy)?, ethyl and methyl esters.
Douglas J, et al.
Organic Syntheses, 56, 59-59 (1977)
2?Hydroxy?3, 5?diiodobenzoic Acid.
Woollett G H and Johnson W W
Organic Syntheses, 14, 52-52 (1943)
Behrooz Darbani
International journal of molecular sciences, 22(11) (2021-06-03)
Gene clusters are becoming promising tools for gene identification. The study reveals the purposive genomic distribution of genes toward higher inheritance rates of intact metabolic pathways/phenotypes and, thereby, higher fitness. The co-localization of co-expressed, co-interacting, and functionally related genes was
L I Wiebe et al.
Drug metabolism and disposition: the biological fate of chemicals, 6(3), 296-302 (1978-05-01)
Butylated hydroxytoluene (BHT) containing the stable isotope 13C was synthesized from 2-[13C]methylpropan-2-ol. A minor constituent of urine following ingestion of BHT-13C by a human volunteer was identified as 3,5-di-(1-[13C]methyl-1-methylethyl)-4-hydroxybenzoic acid, The major metabolite detected was 13C-labeled 5-carboxy-7-(1-carboxy-1-methylethyl)-3,3-dimethyl-2-hydroxy-2,3-dihydrobenzofuran. Detailed spectral analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service