Skip to Content
Merck
All Photos(1)

Key Documents

63627

Supelco

Syringic acid

analytical standard

Synonym(s):

3,5-Dimethoxy-4-hydroxybenzoic acid, 4-Hydroxy-3,5-dimethoxy-benzoic acid, Gallic acid 3,5-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CO2H
CAS Number:
Molecular Weight:
198.17
Beilstein:
2115262
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

mp

205-209 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(cc(OC)c1O)C(O)=O

InChI

1S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10H,1-2H3,(H,11,12)

InChI key

JMSVCTWVEWCHDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Hemmler et al.
Electrophoresis, 39(13), 1645-1653 (2018-03-24)
Comprehensive non-targeted analysis of food products normally requires two complementary chromatographic runs to achieve maximum compound coverage. In this study, we present a sensitive tandem-LC method, which combines RP and HILIC separation in a single run. The setup consists of
Su Jeong Ha et al.
Biochemical pharmacology, 154, 435-445 (2018-06-11)
Validation of nutraceutical and pharmaceutical targets is essential for the prediction of physiological and side effects. Epidemiologic evidence and molecular studies suggest that non-melanoma skin cancer is directly associated with excessive exposure to ultraviolet (UV) radiation. The aim of the
Emin Yilmaz et al.
Journal of food science and technology, 54(10), 3208-3217 (2017-10-05)
Dietary fibers extracted from defatted press meals of orange and grapefruit seeds were used in the production of crackers and the crackers were evaluated for physicochemical, textural, sensory properties, and bioactive compounds. The unexplored fibers (grapes seeds and orange seeds)
B Aydeniz Güneşer et al.
Journal of food science and technology, 56(2), 634-642 (2019-03-25)
This study aims to report the composition of bioactives and volatile aromatic compounds, and determine the descriptive sensory properties of cold-pressed orange seed oil. The effects of oven pre-roasting and microwave pre-roasting of the seeds before cold pressing were compared.
Akinobu Tanaka et al.
Molecules (Basel, Switzerland), 19(6), 8238-8260 (2014-06-20)
Besides being a useful building material, bamboo also is a potential source of bioactive substances. Although some studies have been performed to examine its use in terms of the biological activity, only certain parts of bamboo, especially the leaves or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service