Skip to Content
Merck
All Photos(1)

Key Documents

65009BC

Millipore

SimPlate® Device

BioControl, Quantitative counting method using Binary Detection Technology

Synonym(s):

SimPlate Trays, Sleeve of 20 Trays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

form

solid

shelf life

2‑5 yr

manufacturer/tradename

BioControl Systems 65009-20

storage condition

dry media

application(s)

food and beverages

storage temp.

room temp

General description

SimPlate® Binary Detection Technology combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms resulting in chromogenic or fluorescent changes to the media, providing easy-to-read results. Because SimPlate® relies on these key reactions rather than the formation of a colony, results are available days faster than conventional culture methods.

Application

SimPlate® Device is a quantitative counting method that provides rapid, easy-to-interpret results in food and environmental matrices that are challenging for other rapid methods. SimPlate® tests have been validated by international standards (AOAC/MicroVal)

Features and Benefits

Reduce Costs
  • Faster Results
  • Fewer Dilutions
  • Easy to Prepare
Simplify Procedures
  • Easy-to-Read Results
  • Less Interference
  • Single Plate Results

Components

1 package of 20 trays

Compatibility

AssureSwab sampling device alone will not detect bacterial colonies. It must be used with plating systems such as SimPlate.

Testing Method

Binary Detection Technology®

Other Notes

For use with SimPlate (66002BC, 66003BC, 66004BC, 66005BC, 66006BC, 66007BC, 66008BC, 66009BC)

Legal Information

Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • SimPlate Base/Tray

  • SimPlate Lid

  • Absorbent, Sterile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service